Learn More
N,N'-Diethylethylenediamine, 96%, Thermo Scientific Chemicals
£31.60 - £382.00
Chemical Identifiers
CAS | 111-74-0 |
---|---|
Molecular Formula | C6H16N2 |
Molecular Weight (g/mol) | 116.21 |
MDL Number | MFCD00009033 |
InChI Key | CJKRXEBLWJVYJD-UHFFFAOYSA-N |
Synonym | n,n'-diethylethylenediamine, 1,2-ethanediamine, n,n'-diethyl, 1,2-bis ethylamino ethane, 3,6-diazaoctane, 1,2-ethanediamine, n1,n2-diethyl, n,n'-ethylenediethyldiamine, ethyl 2-ethylamino ethyl amine, ethylenediamine, n,n'-diethyl, n,n'-diethyl-1,2-diaminoethane, dihydrobromide |
PubChem CID | 67105 |
ChEBI | CHEBI:182290 |
IUPAC Name | N,N'-diethylethane-1,2-diamine |
SMILES | CCNCCNCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11498986
|
Thermo Scientific Alfa Aesar
A17113.06 |
5 g |
£31.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11408996
|
Thermo Scientific Alfa Aesar
A17113.14 |
25 g |
£129.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11418996
|
Thermo Scientific Alfa Aesar
A17113.22 |
100 g |
£382.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N,N'-Diethylethylenediamine is used as an internal standard in liquid chromatographic method used for checking the assay of ethambutol. It is utilized for the solvation of lithium hexamethyldisilazide. It acts as a ligand and form coordination complexes such as trans-diaquabis(N,N'-diethylethylenediamine)nickel(II) chloride and copper(II)-N,N-Diethylethylenediamine, which shows thermochromic properties. Further, it is used as a pharmaceutical intermediate used in the synthesis of procainamide hydrochloride, tiapride and cinchocaine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N′-Diethylethylenediamine is used as an internal standard in liquid chromatographic method used for checking the assay of ethambutol. It is utilized for the solvation of lithium hexamethyldisilazide. It acts as a ligand and form coordination complexes such as trans-diaquabis(N,N′-diethylethylenediamine)nickel(II) chloride and copper(II)-N,N-Diethylethylenediamine, which shows thermochromic properties. Further, it is used as a pharmaceutical intermediate used in the synthesis of procainamide hydrochloride, tiapride and cinchocaine.
Solubility
Miscible with water.
Notes
Incompatible with acids, acid chlorides, acid anhydrides, carbon dioxide and strong oxidizing agents.
Chemical Identifiers
111-74-0 | |
116.21 | |
CJKRXEBLWJVYJD-UHFFFAOYSA-N | |
67105 | |
N,N'-diethylethane-1,2-diamine |
C6H16N2 | |
MFCD00009033 | |
n,n'-diethylethylenediamine, 1,2-ethanediamine, n,n'-diethyl, 1,2-bis ethylamino ethane, 3,6-diazaoctane, 1,2-ethanediamine, n1,n2-diethyl, n,n'-ethylenediethyldiamine, ethyl 2-ethylamino ethyl amine, ethylenediamine, n,n'-diethyl, n,n'-diethyl-1,2-diaminoethane, dihydrobromide | |
CHEBI:182290 | |
CCNCCNCC |
Specifications
111-74-0 | |
152°C to 154°C | |
C6H16N2 | |
MFCD00009033 | |
Air Sensitive | |
Miscible with water. | |
CCNCCNCC | |
116.21 | |
CHEBI:182290 | |
96% |
0.811 | |
33°C (92°F) | |
1.4326 | |
UN2685 | |
n,n'-diethylethylenediamine, 1,2-ethanediamine, n,n'-diethyl, 1,2-bis ethylamino ethane, 3,6-diazaoctane, 1,2-ethanediamine, n1,n2-diethyl, n,n'-ethylenediethyldiamine, ethyl 2-ethylamino ethyl amine, ethylenediamine, n,n'-diethyl, n,n'-diethyl-1,2-diaminoethane, dihydrobromide | |
CJKRXEBLWJVYJD-UHFFFAOYSA-N | |
N,N'-diethylethane-1,2-diamine | |
67105 | |
116.21 | |
N,N'-Diethylethylenediamine |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: N,N-DIETHYLETHYLENEDIAMINE
missing translation for 'einecsNumber' : 203-902-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only