Learn More
N,N-Diethylethylenediamine, 98+%, Thermo Scientific Chemicals
£23.10 - £73.70
Chemical Identifiers
CAS | 100-36-7 |
---|---|
Molecular Formula | C6H16N2 |
Molecular Weight (g/mol) | 116.21 |
MDL Number | MFCD00008176 |
InChI Key | UDGSVBYJWHOHNN-UHFFFAOYSA-N |
Synonym | n,n-diethylethylenediamine, 2-diethylamino ethylamine, n,n-diethyl-1,2-ethanediamine, diethylamino ethylamino, n1,n1-diethylethane-1,2-diamine, 2-aminoethyl diethylamine, 2-aminoethyldiethylamine, n,n-diethylethanediamine, ethylenediamine, n,n-diethyl, n,n-diethylethane-1,2-diamine |
PubChem CID | 60993 |
IUPAC Name | N',N'-diethylethane-1,2-diamine |
SMILES | CCN(CC)CCN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10795211
|
Thermo Scientific Acros
150701000 |
100 mL | Glass bottle |
£23.10
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10437360
|
Thermo Scientific Acros
150705000 |
500 mL | Glass bottle |
£73.70
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
100-36-7 | |
116.21 | |
UDGSVBYJWHOHNN-UHFFFAOYSA-N | |
60993 | |
CCN(CC)CCN |
C6H16N2 | |
MFCD00008176 | |
n,n-diethylethylenediamine, 2-diethylamino ethylamine, n,n-diethyl-1,2-ethanediamine, diethylamino ethylamino, n1,n1-diethylethane-1,2-diamine, 2-aminoethyl diethylamine, 2-aminoethyldiethylamine, n,n-diethylethanediamine, ethylenediamine, n,n-diethyl, n,n-diethylethane-1,2-diamine | |
N',N'-diethylethane-1,2-diamine |
Spécification
100-36-7 | |
100.0 | |
0.8200g/mL | |
30°C | |
98+% | |
(C2H5)2NCH2CH2NH2 | |
MFCD00008176 | |
0.82 | |
Solubility in water: miscible. Other solubilities: soluble in ether,ethanol,toluene,ccl4 | |
CCN(CC)CCN | |
116.21 | |
116.21 | |
Liquid |
98.0 | |
Colorless to Yellow | |
145°C to 147°C | |
Authentic | |
C6H16N2 | |
1.4350 to 1.437 | |
04, 251 | |
n,n-diethylethylenediamine, 2-diethylamino ethylamine, n,n-diethyl-1,2-ethanediamine, diethylamino ethylamino, n1,n1-diethylethane-1,2-diamine, 2-aminoethyl diethylamine, 2-aminoethyldiethylamine, n,n-diethylethanediamine, ethylenediamine, n,n-diethyl, n,n-diethylethane-1,2-diamine | |
UDGSVBYJWHOHNN-UHFFFAOYSA-N | |
N',N'-diethylethane-1,2-diamine | |
60993 | |
98+% | |
N, N-Diethylethylenediamine, 98+% |
Sécurité et traitement
GHS H Statement
Flammable liquid and vapor.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-844-7
missing translation for 'rtecsNumber' : KV3500000
missing translation for 'tsca' : TSCA
- N,N-Diethylethylenediamine
Mention d'avertissement
- Danger
Catégorie de danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
Mention de danger
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H311-Toxic in contact with skin.
- H226-Flammable liquid and vapour.
Conseils de prudence
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only