Learn More
N,N-Diethylpropargylamine, 97%, Thermo Scientific Chemicals
£26.90 - £307.00
Chemical Identifiers
CAS | 4079-68-9 |
---|---|
Molecular Formula | C7H13N |
Molecular Weight (g/mol) | 111.18 |
InChI Key | JZJXKEWVUBVOEH-UHFFFAOYSA-N |
PubChem CID | 20010 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15831012
|
Thermo Scientific Acros
455580050 |
5 g |
£26.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15841012
|
Thermo Scientific Acros
455580250 |
25 g |
£85.90
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15851012
|
Thermo Scientific Acros
455581000 |
100 g |
£307.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4079-68-9 | |
118°C to 120°C | |
96% min. (GC) | |
0.811 | |
111.18 | |
111.18 | |
N,N-Diethylpropargylamine |
0.8110g/mL | |
20°C | |
C7H13N | |
JZJXKEWVUBVOEH-UHFFFAOYSA-N | |
20010 | |
97% |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapour.
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 223-804-5
RUO – Research Use Only