missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diisobutyl-1,6-hexanediamine, 97%, Thermo Scientific™

Product Code. 11440107
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16121-92-9
Molecular Formula C14H32N2
Molecular Weight (g/mol) 228.424
MDL Number MFCD00048339
InChI Key LTRQCWUIJWVNNK-UHFFFAOYSA-N
Synonym n,n'-diisobutyl-1,6-hexanediamine, 1,6-hexanediamine, n,n'-bis 2-methylpropyl, n,n'-diisobutylhexanediamine, acmc-1bs6j, diisobutyl-hexamethylendiamin, n,n/'-diisobutylhexanediamine, n,n'-di isobutyl hexamethylenediamine, n,n'-diisobutyl-1,6-hexane diamine, n,n'-bis 2-methylpropyl hexane-1,6-diamine
PubChem CID 177722
IUPAC Name N,N'-bis(2-methylpropyl)hexane-1,6-diamine
SMILES CC(C)CNCCCCCCNCC(C)C

Specifications

Quantity 5 g
Formula Weight 228.41
Percent Purity 97%
Chemical Name or Material N,N'-Diisobutyl-1,6-hexanediamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'