missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diisopropylcarbodiimide, 99%, Thermo Scientific Chemicals

Alfa Aesar N,N'-Diisopropylcarbodiimide, 99%, Quantity: 50g, Boiling Point: 145 deg.C to 148 deg.C, Molecular Weight: 126.203, Percent Purity: 99%, Beilstein: 878281, CAS: 693-13-0, ChEBI: CHEBI:53092, Density: 0.81, DOT Information: Hazard Class: 6.1; Packaging Group: I, EINECS Number: 211-743-7

£39.09 - £434.16

Chemical Identifiers

CAS 693-13-0
Molecular Formula C7H14N2
Molecular Weight (g/mol) 126.203
MDL Number MFCD00065689
InChI Key BDNKZNFMNDZQMI-UHFFFAOYSA-N
Synonym n,n'-diisopropylcarbodiimide, diisopropylcarbodiimide, 1,3-diisopropylcarbodiimide, n,n-diisopropylcarbodiimide, carbodiimide, diisopropyl, dipcdi, dic, 2-propanamine, n,n'-methanetetraylbis, diisopropylmethanediimine, n,n'-methanediylidenebis propan-2-amine
PubChem CID 12734
ChEBI CHEBI:53092
IUPAC Name N,N'-di(propan-2-yl)methanediimine
SMILES CC(C)N=C=NC(C)C
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11424230
View Documents
Thermo Scientific Alfa Aesar
A19292.09
10 g
£39.09
10g
Please to purchase this item. Need a web account? Register with us today!
11409556
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A19292.14
25 g
£78.49
25g
Please to purchase this item. Need a web account? Register with us today!
11434230
View Documents
Thermo Scientific Alfa Aesar
A19292.18
50 g
£130.65
50g
Please to purchase this item. Need a web account? Register with us today!
11444230
View Documents
Thermo Scientific Alfa Aesar
A19292.30
250 g
£434.16
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

N,N'-Diisopropylcarbodiimide is used as a reagent in synthetic organic chemistry. It serves as a chemical intermediate and as a stabilizer for Sarin (chemical weapon). It is also used in the synthesis of peptide and nucleic acid. Further, it is used as an antineoplastic and involved in the treatment of malignant melanoma and sarcomas. In addition to this, it is used in the synthesis of acid anhydride, aldehyde, ketone and isocyanate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Diisopropylcarbodiimide is used as a reagent in synthetic organic chemistry. It serves as a chemical intermediate and as a stabilizer for Sarin (chemical weapon). It is also used in the synthesis of peptide and nucleic acid. Further, it is used as an antineoplastic and involved in the treatment of malignant melanoma and sarcomas. In addition to this, it is used in the synthesis of acid anhydride, aldehyde, ketone and isocyanate.

Solubility
Soluble in chloroform, methylene chloride, acetonitrile, dioxane, dimethylformamide andtetrahydrofuran.

Notes
Moisture sensitive. Store in a cool place Incompatible with oxidizing agents and acids.
Specifications

Chemical Identifiers

693-13-0
126.203
BDNKZNFMNDZQMI-UHFFFAOYSA-N
12734
N,N'-di(propan-2-yl)methanediimine
C7H14N2
MFCD00065689
n,n'-diisopropylcarbodiimide, diisopropylcarbodiimide, 1,3-diisopropylcarbodiimide, n,n-diisopropylcarbodiimide, carbodiimide, diisopropyl, dipcdi, dic, 2-propanamine, n,n'-methanetetraylbis, diisopropylmethanediimine, n,n'-methanediylidenebis propan-2-amine
CHEBI:53092
CC(C)N=C=NC(C)C

Specifications

693-13-0
145°C to 148°C
Stench
1.433
UN2929
Moisture sensitive
Soluble in chloroform,methylene chloride,acetonitrile,dioxane,dimethylformamide andtetrahydrofuran.
CC(C)N=C=NC(C)C
126.203
CHEBI:53092
99%
0.81
33°C (91°F)
C7H14N2
MFCD00065689
878281
n,n'-diisopropylcarbodiimide, diisopropylcarbodiimide, 1,3-diisopropylcarbodiimide, n,n-diisopropylcarbodiimide, carbodiimide, diisopropyl, dipcdi, dic, 2-propanamine, n,n'-methanetetraylbis, diisopropylmethanediimine, n,n'-methanediylidenebis propan-2-amine
BDNKZNFMNDZQMI-UHFFFAOYSA-N
N,N'-di(propan-2-yl)methanediimine
12734
126.2
N,N'-Diisopropylcarbodiimide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H330-H334-H318-H226-H315-H317-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Flammable liquid and vapour.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P272-P280-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P333+P313-P363-P370+P378q-P501c

H226-H315-H317-H318-H330-H334-H335

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: I

missing translation for 'einecsNumber' : 211-743-7

missing translation for 'rtecsNumber' : FF2175000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Product Identifier
  • N,N'-Diisopropylcarbodiimide

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Diisopropylcarbodiimide

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only