missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diisopropylcarbodiimide, 99%, Thermo Scientific Chemicals

Product Code. 10193531
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-13-0
Molecular Formula C7H14N2
Molecular Weight (g/mol) 126.2
MDL Number MFCD00065689
InChI Key BDNKZNFMNDZQMI-UHFFFAOYSA-N
Synonym n,n'-diisopropylcarbodiimide, diisopropylcarbodiimide, 1,3-diisopropylcarbodiimide, n,n-diisopropylcarbodiimide, carbodiimide, diisopropyl, dipcdi, dic, 2-propanamine, n,n'-methanetetraylbis, diisopropylmethanediimine, n,n'-methanediylidenebis propan-2-amine
PubChem CID 12734
ChEBI CHEBI:53092
IUPAC Name N,N'-di(propan-2-yl)methanediimine
SMILES CC(C)N=C=NC(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8100g/mL
Boiling Point 145.0°C to 148.0°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHN=C=NCH(CH3)2
Refractive Index 1.4320 to 1.4350
Quantity 25 g
Merck Index 15,3218
Specific Gravity 0.81
Solubility Information Solubility in water: decomposes
Formula Weight 126.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N, N'-Diisopropylcarbodiimide, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'