missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diisopropylethylamine, 99+%, Thermo Scientific Chemicals

Product Code. 10008129
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic Bottle
Plastic Can
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7087-68-5
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.24
MDL Number MFCD00008868
InChI Key JGFZNNIVVJXRND-UHFFFAOYSA-N
Synonym n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine
PubChem CID 81531
IUPAC Name N-ethyl-N-propan-2-ylpropan-2-amine
SMILES CCN(C(C)C)C(C)C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -46.0°C
Color Colorless to Yellow
Density 0.7560g/mL
Boiling Point 127.0°C
Flash Point 6°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic Bottle
Linear Formula [(CH3)2CH]2NC2H5
Refractive Index 1.4123 to 1.4143
Quantity 2.5 kg
Beilstein 04,IV,511
Fieser 01,371; 04,230; 07,148; 10,151; 12,277; 13,121; 16,176
Merck Index 15,3219
Specific Gravity 0.756
Solubility Information Solubility in water: 3.9g/L (20°C).
Viscosity 0.88 mm2/s (20°C)
Formula Weight 129.24
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material N,N-Diisopropylethylamine, 98+%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Diisopropylethylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.