missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dimethyl-1,3-propanediamine, 97%, Thermo Scientific Chemicals

Product Code. 11418726
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Employed as a pharmaceutical intermediate. It is an important raw material used in organic synthesis, agrochemicals and dye stuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a pharmaceutical intermediate. It is an important raw material used in organic synthesis, agrochemicals and dye stuffs.

Solubility
Fully miscible in water, 1e+006 mg/L @ 25°C (est).

Notes
Air sensitive. Store away from oxidizing agents, carbon dioxide and air. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-33-1
Molecular Formula C5H16N2
Molecular Weight (g/mol) 104.20
MDL Number MFCD00008292
InChI Key UQUPIHHYKUEXQD-UHFFFAOYSA-P
Synonym n,n'-dimethyl-1,3-propanediamine, n1,n3-dimethylpropane-1,3-diamine, 1,3-propanediamine, n,n'-dimethyl, 1,3-bis methylamino propane, methyl 3-methylamino propyl amine, 1,3-propanediamine, n1,n3-dimethyl, n,n'-dimethyltrimethylenediamine, n1,n3-dimethyl-1,3-propanediamine, n-3-methylaminopropyl-n-methylamine, n,n'-dimethylpropan-1,3-diamin
PubChem CID 66978
IUPAC Name N,N'-dimethylpropane-1,3-diamine
SMILES C[NH2+]CCC[NH2+]C

Specifications

Density 0.817
Boiling Point 143°C to 146°C
Flash Point 32°C (90°F)
Odor Amine-like
Quantity 5 g
UN Number UN2733
Sensitivity Air sensitive; Hygroscopic
Solubility Information Fully miscible in water,1e+006mg/L @ 25°C (est).
Formula Weight 102.18
Percent Purity 97%
Chemical Name or Material N,N'-Dimethyl-1,3-propanediamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'