missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dimethyl-1,3-propanediamine, 97%, Thermo Scientific Chemicals

£80.00 - £261.30

Chemical Identifiers

CAS 111-33-1
Molecular Formula C5H16N2
Molecular Weight (g/mol) 104.20
MDL Number MFCD00008292
InChI Key UQUPIHHYKUEXQD-UHFFFAOYSA-P
Synonym n,n'-dimethyl-1,3-propanediamine, n1,n3-dimethylpropane-1,3-diamine, 1,3-propanediamine, n,n'-dimethyl, 1,3-bis methylamino propane, methyl 3-methylamino propyl amine, 1,3-propanediamine, n1,n3-dimethyl, n,n'-dimethyltrimethylenediamine, n1,n3-dimethyl-1,3-propanediamine, n-3-methylaminopropyl-n-methylamine, n,n'-dimethylpropan-1,3-diamin
PubChem CID 66978
IUPAC Name N,N'-dimethylpropane-1,3-diamine
SMILES C[NH2+]CCC[NH2+]C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10751921
View Documents Promotion Details
Thermo Scientific Acros
314830050
5 g Glass bottle
£80.00
5g
Please to purchase this item. Need a web account? Register with us today!
10454215
View Documents Promotion Details
Thermo Scientific Acros
314830250
25 g Glass bottle
£261.30
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

111-33-1
104.20
UQUPIHHYKUEXQD-UHFFFAOYSA-P
66978
C[NH2+]CCC[NH2+]C
C5H16N2
MFCD00008292
n,n'-dimethyl-1,3-propanediamine, n1,n3-dimethylpropane-1,3-diamine, 1,3-propanediamine, n,n'-dimethyl, 1,3-bis methylamino propane, methyl 3-methylamino propyl amine, 1,3-propanediamine, n1,n3-dimethyl, n,n'-dimethyltrimethylenediamine, n1,n3-dimethyl-1,3-propanediamine, n-3-methylaminopropyl-n-methylamine, n,n'-dimethylpropan-1,3-diamin
N,N'-dimethylpropane-1,3-diamine

Specifications

111-33-1
100.0
0.8100g/mL
20°C
96% min. (GC)
CH3NH(CH2)3NHCH3
MFCD00008292
n,n'-dimethyl-1,3-propanediamine, n1,n3-dimethylpropane-1,3-diamine, 1,3-propanediamine, n,n'-dimethyl, 1,3-bis methylamino propane, methyl 3-methylamino propyl amine, 1,3-propanediamine, n1,n3-dimethyl, n,n'-dimethyltrimethylenediamine, n1,n3-dimethyl-1,3-propanediamine, n-3-methylaminopropyl-n-methylamine, n,n'-dimethylpropan-1,3-diamin
C[NH2+]CCC[NH2+]C
104.20
102.18
Liquid
96.0
Colorless
145.0°C
Authentic
C5H16N2
1.4375 to 1.4395
0.81
UQUPIHHYKUEXQD-UHFFFAOYSA-P
N,N'-dimethylpropane-1,3-diamine
66978
97%
N,N′-Dimethyl-1,3-propanediamine, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 203-859-1

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only