missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethyl-p-phenylenediamine dihydrochloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
It is a stain used in microscopy. N,N-Dimethyl-p-phenylenediamine dihydrochloride is also suitable for use in the peroxidase test. It is a compound that is normally used to measure the antioxidant potential.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a stain used in microscopy. N,N-Dimethyl-p-phenylenediamine dihydrochloride is also suitable for use in the peroxidase test. It is a compound that is normally used to measure the antioxidant potential.
Solubility
Soluble in water
Notes
Store away from oxidizing agents, humidity and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from light. Store under inert gas.
It is a stain used in microscopy. N,N-Dimethyl-p-phenylenediamine dihydrochloride is also suitable for use in the peroxidase test. It is a compound that is normally used to measure the antioxidant potential.
Solubility
Soluble in water
Notes
Store away from oxidizing agents, humidity and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from light. Store under inert gas.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 212°C (decomposition) |
Quantity | 25 g |
UN Number | UN2811 |
Sensitivity | Hygroscopic; Light sensitive |
Solubility Information | Soluble in water |
IUPAC Name | 4-N,4-N-dimethylbenzene-1,4-diamine;dihydrochloride |
Formula Weight | 209.12 |
Percent Purity | 98% |
Chemical Name or Material | N,N-Dimethyl-p-phenylenediamine dihydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'