missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-p-toluidine, 99%

Product Code. 10543884 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10543884

Brand: Thermo Scientific Acros 116900025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-97-8 | C9H13N | 135.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-97-8
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00008316
InChI Key GYVGXEWAOAAJEU-UHFFFAOYSA-N
Synonym n,n-dimethyl-p-toluidine, dimethyl-p-toluidine, benzenamine, n,n,4-trimethyl, n,n-dimethyl-4-methylaniline, n,n-dimethyl-para-toluidine, 4,n,n-trimethylaniline, dimethyl-4-toluidine, p-dimethylamino toluene, n,n-dimethyl-p-tolylamine, n,n,4-trimethylbenzenamine
PubChem CID 7471
IUPAC Name N,N,4-trimethylaniline
SMILES CN(C)C1=CC=C(C)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 0.9300g/mL
Boiling Point 211.0°C
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4N(CH3)2
Refractive Index 1.5450 to 1.5470
Quantity 2.5 kg
Beilstein 12, 902
Specific Gravity 0.93
Solubility Information Solubility in water: immiscible
Formula Weight 135.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N, N-Dimethyl-p-toluidine, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethyl-p-toluidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H317-May cause an allergic skin reaction.
  • H330-Fatal if inhaled.
  • H350-May cause cancer.
  • H361-Suspected of damaging fertility or the unborn child.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.