Learn More
N,N-Dimethyl-p-toluidine, 99%, Thermo Scientific Chemicals
£27.10 - £308.00
Chemical Identifiers
CAS | 99-97-8 |
---|---|
Molecular Formula | C9H13N |
Molecular Weight (g/mol) | 135.21 |
MDL Number | MFCD00008316 |
InChI Key | GYVGXEWAOAAJEU-UHFFFAOYSA-N |
Synonym | n,n-dimethyl-p-toluidine, dimethyl-p-toluidine, benzenamine, n,n,4-trimethyl, n,n-dimethyl-4-methylaniline, n,n-dimethyl-para-toluidine, 4,n,n-trimethylaniline, dimethyl-4-toluidine, p-dimethylamino toluene, n,n-dimethyl-p-tolylamine, n,n,4-trimethylbenzenamine |
PubChem CID | 7471 |
IUPAC Name | N,N,4-trimethylaniline |
SMILES | CN(C)C1=CC=C(C)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11404034
|
Thermo Scientific Alfa Aesar
A11988.22 |
100 g |
£27.10
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11414034
|
Thermo Scientific Alfa Aesar
A11988.36 |
500 g |
£80.90
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11424034
|
Thermo Scientific Alfa Aesar
A11988.0E |
2500 g |
£308.00
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N,N-Dimethyl-p-toluidine is used as a polymerization catalyst for polyesters, acrylate and epoxy resins. It is also used as a hardener for dental cements and in adhesives. It serves as an intermediate for photographic chemicals, in industrial glues, in artificial fingernail preparations, colorants, pharmaceuticals. It reacts with vinyl ether in the presence of copper(II) chloride gives tetrahydroquinolines. Further, it is used to accelerate polymerization of ethyl methacrylate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N-Dimethyl-p-toluidine is used as a polymerization catalyst for polyesters, acrylate and epoxy resins. It is also used as a hardener for dental cements and in adhesives. It serves as an intermediate for photographic chemicals, in industrial glues, in artificial fingernail preparations, colorants, pharmaceuticals. It reacts with vinyl ether in the presence of copper(II) chloride gives tetrahydroquinolines. Further, it is used to accelerate polymerization of ethyl methacrylate.
Solubility
Miscible with alcohol, ether and chloroform. Immiscible with water.
Notes
Incompatible with strong oxidizing agents. Store in a cool place.
Chemical Identifiers
99-97-8 | |
135.21 | |
GYVGXEWAOAAJEU-UHFFFAOYSA-N | |
7471 | |
CN(C)C1=CC=C(C)C=C1 |
C9H13N | |
MFCD00008316 | |
n,n-dimethyl-p-toluidine, dimethyl-p-toluidine, benzenamine, n,n,4-trimethyl, n,n-dimethyl-4-methylaniline, n,n-dimethyl-para-toluidine, 4,n,n-trimethylaniline, dimethyl-4-toluidine, p-dimethylamino toluene, n,n-dimethyl-p-tolylamine, n,n,4-trimethylbenzenamine | |
N,N,4-trimethylaniline |
Specifications
99-97-8 | |
0.937 | |
83°C (181°F) | |
1.546 | |
UN1708 | |
n,n-dimethyl-p-toluidine, dimethyl-p-toluidine, benzenamine, n,n,4-trimethyl, n,n-dimethyl-4-methylaniline, n,n-dimethyl-para-toluidine, 4,n,n-trimethylaniline, dimethyl-4-toluidine, p-dimethylamino toluene, n,n-dimethyl-p-tolylamine, n,n,4-trimethylbenzenamine | |
GYVGXEWAOAAJEU-UHFFFAOYSA-N | |
N,N,4-trimethylaniline | |
7471 | |
99% |
-25°C | |
210°C to 211°C | |
C9H13N | |
MFCD00008316 | |
774409 | |
Miscible with alcohol,ether and chloroform. Immiscible with water. | |
CN(C)C1=CC=C(C)C=C1 | |
135.21 | |
135.21 | |
N,N-Dimethyl-p-toluidine |
Safety and Handling
GHS H Statement
H301-H311-H331-H373-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
P210-P235-P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P370+P378q-P501c
H227-H301+H311+H331-H373
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 202-805-4
missing translation for 'rtecsNumber' : XU5803000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only