Learn More
N,N-Dimethylacetamide, 99%, Thermo Scientific Chemicals
£20.00 - £71.56
Chemical Identifiers
CAS | 127-19-5 |
---|---|
Molecular Formula | C4H9NO |
Molecular Weight (g/mol) | 87.12 |
MDL Number | MFCD00008686 |
InChI Key | FXHOOIRPVKKKFG-UHFFFAOYSA-N |
Synonym | dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamineShow More |
PubChem CID | 31374 |
ChEBI | CHEBI:84254 |
IUPAC Name | N,N-dimethylacetamide |
SMILES | CN(C)C(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11484313
|
Thermo Scientific Alfa Aesar
A10924.AP |
500 mL |
£20.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11494313
|
Thermo Scientific Alfa Aesar
A10924.0F |
2500 mL |
£71.56
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N,N-Dimethylacetamide is used as a polar solvent in organic synthesis as well as in reactions involving strong bases such as sodium hydroxide. It is also used as a solvent for fibers like polyacrylonitrile, spandex, as an excipient in drugs viz. vumon, busulfex and in the adhesive industry. It finds application as a reaction medium in the production of pharmaceuticals, agrochemicals, dyes and plasticizers. It is widely used in polymer industry due to its high solving power for high molecular-weight polymers and synthetic resins. It plays an important role as a catalyst in various reactions viz. cyclization, halogenation, cynidation, alkylation and dehydrogenation and increases the yield of main products. It is also used as an extracting agent for oil and gases, in paint removers, in the production of photo-resist stripping compounds and as booster solvent in coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N-Dimethylacetamide is used as a polar solvent in organic synthesis as well as in reactions involving strong bases such as sodium hydroxide. It is also used as a solvent for fibers like polyacrylonitrile, spandex, as an excipient in drugs viz. vumon, busulfex and in the adhesive industry. It finds application as a reaction medium in the production of pharmaceuticals, agrochemicals, dyes and plasticizers. It is widely used in polymer industry due to its high solving power for high molecular-weight polymers and synthetic resins. It plays an important role as a catalyst in various reactions viz. cyclization, halogenation, cynidation, alkylation and dehydrogenation and increases the yield of main products. It is also used as an extracting agent for oil and gases, in paint removers, in the production of photo-resist stripping compounds and as booster solvent in coatings.
Solubility
Miscible with water, benzene, alcohol, acetone, ether, esters and ketones.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Chemical Identifiers
127-19-5 | |
87.12 | |
FXHOOIRPVKKKFG-UHFFFAOYSA-N | |
31374 | |
N,N-dimethylacetamide |
C4H9NO | |
MFCD00008686 | |
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamineShow More | |
CHEBI:84254 | |
CN(C)C(C)=O |
Specifications
127-19-5 | |
0.94 | |
70°C (158°F) | |
C4H9NO | |
1.4385 | |
1737614 | |
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine | |
FXHOOIRPVKKKFG-UHFFFAOYSA-N | |
N,N-dimethylacetamide | |
31374 | |
87.12 | |
N,N-Dimethylacetamide |
-20°C | |
165°C to 166°C | |
Ammonia-like | |
CH3CON(CH3)2 | |
MFCD00008686 | |
14,3227 | |
Miscible with water,benzene,alcohol,acetone,ether,esters and ketones. | |
CN(C)C(C)=O | |
87.12 | |
CHEBI:84254 | |
99% |
Safety and Handling
GHS H Statement
H360-H312-H332-H227
May damage fertility or the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.
P201-P202-P210-P261-P271-P281-P302+P352-P304+P340-P308+P313-P312-P363-P370+P378q-P501c
H227-H312+H332-H360D
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 204-826-4
missing translation for 'rtecsNumber' : AB7700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- N,N-Dimethylacetamide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
Hazard Statement
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H360D-May damage the unborn child.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : N,N-Dimethylacetamide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.