missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetamide, 99+%, for spectroscopy, Thermo Scientific Chemicals

£24.72 - £119.60

Chemical Identifiers

CAS 127-19-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00008686
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
PubChem CID 31374
ChEBI CHEBI:84254
IUPAC Name N,N-dimethylacetamide
SMILES CN(C)C(C)=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10762591
missing translation for 'viewDocuments'
Thermo Scientific Acros
167782500
250 mL Glass Bottle
£24.72
250mL
Please to purchase this item. Need a web account? Register with us today!
10012220
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Acros
167780010
1 L Glass Bottle
£58.99
1 L
Please to purchase this item. Need a web account? Register with us today!
10522934
missing translation for 'viewDocuments'
Thermo Scientific Acros
167780025
2.5 L Glass Bottle
£119.60
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
Specifications

Chemical Identifiers

127-19-5
87.12
FXHOOIRPVKKKFG-UHFFFAOYSA-N
31374
N,N-dimethylacetamide
C4H9NO
MFCD00008686
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
CHEBI:84254
CN(C)C(C)=O

Specifications

-20.0°C
127-19-5
100.0
164.0°C to 166.0°C
Liquid
99+%
0.937
CH3CON(CH3)2
1.02 mPa.s (20°C)
0.05% max.
15, 3248
Solubility in water: soluble. Other solubilities: soluble in acetone, diethyl ether, benzene,, chloroform, ethanol and toluene
CN(C)C(C)=O
87.12
CHEBI:84254
≥99%
N, N-Dimethylacetamide
0.01 max. at 360nm, 0.03 max. at 320nm, 0.05 max. at 310nm, 0.15 max. at 290nm, 0.3 max. at 280nm, 1 max. at 270nm
99.0
0.9370g/mL
70°C
1.4370 to 1.4390
0.0005% max.
C4H9NO
MFCD00008686
04, 59
01,270; 02,144; 04,165
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
FXHOOIRPVKKKFG-UHFFFAOYSA-N
N,N-dimethylacetamide
31374
87.12
Spectroscopy
Videos
Safety and Handling

Safety and Handling

GHS P Statement
May damage the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
Documents

Documents

Certificates

RUO – Research Use Only