missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylacetamide, BAKER ANALYZED™ Reagent, J.T. Baker™
426.55 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
Specifications
Quantity | 500 mL |
Flash Point | 70.00°C |
Packaging | Narrow Mouth Amber Glass Bottle |
Specific Gravity | 1L = 0.94kg |
Formula Weight | 87.12 |
Grade | BAKER ANALYZED™ Reagent |
Chemical Name or Material | N,N-Dimethylacetamide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'