missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetamide dimethyl acetal, 97%, stabilized, Thermo Scientific Chemicals

£147.74 - £484.41

Chemical Identifiers

CAS 18871-66-4
Molecular Formula C6H15NO2
Molecular Weight (g/mol) 133.19
MDL Number MFCD00008476
InChI Key FBZVZUSVGKOWHG-UHFFFAOYSA-N
Synonym n,n-dimethylacetamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylethylamine, 1,1-dimethoxyethyl dimethylamine, 1,1-dimethoxyethyl dimethyl amine, 1,1-dimethoxy-ethyl-dimethyl-amine, ethanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylacetamide dimethylacetal, dimethylacetamide dimethyl acetal, n,n-dimethylacetamide dimethyl acetal, stabilized with methanol, zlchem 332
PubChem CID 87835
IUPAC Name 1,1-dimethoxy-N,N-dimethylethanamine
SMILES CC(N(C)C)(OC)OC
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10270853
SDS View Documents
Thermo Scientific Acros
264130250
25 g Glass bottle
£147.74
25g
Please to purchase this item. Need a web account? Register with us today!
10701925
SDS View Documents
Thermo Scientific Acros
264131000
100 g Glass bottle
£484.41
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and eas

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 242-641-

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N-Dimethylacetamide dimethyl acetal, 97%, stabilized, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.