Learn More
N,N-Dimethylacrylamide, 99%, stabilized with 500 ppm MEHQ, Thermo Scientific Chemicals
£40.50 - £170.85
Chemical Identifiers
CAS | 2680-03-7 |
---|---|
Molecular Formula | C5H9NO |
Molecular Weight (g/mol) | 99.13 |
InChI Key | YLGYACDQVQQZSW-UHFFFAOYSA-N |
Synonym | n,n-dimethylacrylamide, 2-propenamide, n,n-dimethyl, acrylamide, n,n-dimethyl, acylamide, n,n-dimethyl, n,n-dimethyl-acrylamide, n,n-dimethyl-2-propenamide, dimethylamid kyseliny akrylove, nn-dimethylacrylamide, unii-as46jk7q6i, propenamide, n,n-dimethyl |
PubChem CID | 17587 |
IUPAC Name | N,N-dimethylprop-2-enamide |
SMILES | CN(C)C(=O)C=C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11379671
|
Thermo Scientific Acros
432021000 |
100 mL | Glass bottle |
£40.50
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11915481
|
Thermo Scientific Acros
432025000 |
500 mL | Glass bottle |
£170.85
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2680-03-7 | |
99.13 | |
n,n-dimethylacrylamide, 2-propenamide, n,n-dimethyl, acrylamide, n,n-dimethyl, acylamide, n,n-dimethyl, n,n-dimethyl-acrylamide, n,n-dimethyl-2-propenamide, dimethylamid kyseliny akrylove, nn-dimethylacrylamide, unii-as46jk7q6i, propenamide, n,n-dimethyl | |
N,N-dimethylprop-2-enamide |
C5H9NO | |
YLGYACDQVQQZSW-UHFFFAOYSA-N | |
17587 | |
CN(C)C(=O)C=C |
Specifications
2680-03-7 | |
100.0 | |
0.9620g/mL | |
70°C | |
98.5% min. (GC) | |
1.4720 to 1.4740 (20°C, 589nm) | |
n,n-dimethylacrylamide, 2-propenamide, n,n-dimethyl, acrylamide, n,n-dimethyl, acylamide, n,n-dimethyl, n,n-dimethyl-acrylamide, n,n-dimethyl-2-propenamide, dimethylamid kyseliny akrylove, nn-dimethylacrylamide, unii-as46jk7q6i, propenamide, n,n-dimethyl | |
YLGYACDQVQQZSW-UHFFFAOYSA-N | |
N,N-dimethylprop-2-enamide | |
99.13 | |
99.13 | |
Liquid |
98.5 | |
Colorless | |
80.0°C (20.0 mmHg) | |
Authentic | |
C5H9NO | |
0.962 | |
Solubility in water: slightly soluble. | |
CN(C)C(=O)C=C | |
500ppm (approximately) | |
17587 | |
99% | |
N, N-Dimethylacrylamide, Stabilized |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 220-237-5
- N,N-Dimethylacrylamide, stabilized with 500 ppm MEHQ
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H311-Toxic in contact with skin.
- H301-Toxic if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only