missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylaniline, 99%, Thermo Scientific Chemicals

£24.42 - £233.16

Chemical Identifiers

CAS 121-69-7
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00008304
InChI Key JLTDJTHDQAWBAV-UHFFFAOYSA-N
Synonym dimethylaniline, dimethylphenylamine, n,n-dimethylbenzenamine, benzenamine, n,n-dimethyl, dimethylamino benzene, n,n-dimethylphenylamine, n,n-dimethylbenzeneamine, dimethylaminobenzene, dwumetyloanilina, n,n-dimethylamino benzene
PubChem CID 949
ChEBI CHEBI:16269
IUPAC Name N,N-dimethylaniline
SMILES CN(C)C1=CC=CC=C1
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10356600
View Documents
Thermo Scientific Acros
115920050
5 mL Glass Bottle
£24.42
5mL
Please to purchase this item. Need a web account? Register with us today!
10336650
View Documents Promotion Details
Thermo Scientific Acros
1 L Glass Bottle
£39.30
1 L
Please to purchase this item. Need a web account? Register with us today!
10580571
View Documents
Thermo Scientific Acros
115920025
2.5 L Glass Bottle
£84.62
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10125633
View Documents
Thermo Scientific Acros
115920100
10 L Metal drum
£233.16
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

121-69-7
121.18
JLTDJTHDQAWBAV-UHFFFAOYSA-N
949
N,N-dimethylaniline
C8H11N
MFCD00008304
dimethylaniline, dimethylphenylamine, n,n-dimethylbenzenamine, benzenamine, n,n-dimethyl, dimethylamino benzene, n,n-dimethylphenylamine, n,n-dimethylbenzeneamine, dimethylaminobenzene, dwumetyloanilina, n,n-dimethylamino benzene
CHEBI:16269
CN(C)C1=CC=CC=C1

Specifications

121-69-7
100.0
Yellow
193.0°C to 194.0°C
Authentic
C8H11N
1.5570 to 1.5590
01,274; 02,147
0.95
Solubility in water: 1g/L (20°C). Other solubilities: freely soluble in alcohol, ether, chloroform, acet, carbon tetrachloride
CN(C)C1=CC=CC=C1
121.18
CHEBI:16269
99%
N, N-Dimethylaniline
98.5
1.5°C to 2.5°C
0.9500g/mL
63°C
98.5% min. (GC)
C6H5N(CH3)2
MFCD00008304
15, 3256
dimethylaniline, dimethylphenylamine, n,n-dimethylbenzenamine, benzenamine, n,n-dimethyl, dimethylamino benzene, n,n-dimethylphenylamine, n,n-dimethylbenzeneamine, dimethylaminobenzene, dwumetyloanilina, n,n-dimethylamino benzene
JLTDJTHDQAWBAV-UHFFFAOYSA-N
N,N-dimethylaniline
949
121.18
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Toxic in contact with skin.
Suspected of causing cancer.
Toxic if inhaled.
Toxic if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 204-493-5

Product Identifier
  • N,N-Dimethylaniline

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 2

Hazard Statement
  • H351-Suspected of causing cancer.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only