Learn More
N,N-Dimethylethanolamine, 99%, Thermo Scientific Chemicals
£21.20 - £246.00
Chemical Identifiers
CAS | 108-01-0 |
---|---|
Molecular Formula | C4H11NO |
Molecular Weight (g/mol) | 89.14 |
MDL Number | MFCD00002846 |
InChI Key | UEEJHVSXFDXPFK-UHFFFAOYSA-N |
Synonym | 2-dimethylamino ethanol, n,n-dimethylethanolamine, deanol, dimethylaminoethanol, dimethylethanolamine, norcholine, dmae, dmea, bimanol, liparon |
PubChem CID | 7902 |
ChEBI | CHEBI:271436 |
IUPAC Name | 2-(dimethylamino)ethanol |
SMILES | CN(C)CCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10132480
|
Thermo Scientific Acros
116182500 |
250 mL | Glass bottle |
£21.20
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10112480
|
Thermo Scientific Acros
116180010 |
1 L | Glass bottle |
£38.40
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10358793
|
Thermo Scientific
116180051 |
5 L | Plastic drum |
£121.00
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10594683
|
Thermo Scientific Acros
116180250 |
25 L | Metal Drum |
£246.00
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
108-01-0 | |
89.14 | |
UEEJHVSXFDXPFK-UHFFFAOYSA-N | |
7902 | |
2-(dimethylamino)ethanol |
C4H11NO | |
MFCD00002846 | |
2-dimethylamino ethanol, n,n-dimethylethanolamine, deanol, dimethylaminoethanol, dimethylethanolamine, norcholine, dmae, dmea, bimanol, liparon | |
CHEBI:271436 | |
CN(C)CCO |
Specifications
108-01-0 | |
100.0 | |
Colorless to Yellow | |
139.0°C | |
Authentic | |
C4H11NO | |
1.4290 to 1.4300 | |
04, 276 | |
0.888 | |
Solubility in water: miscible. Other solubilities: miscible with alcohol and ether | |
CN(C)CCO | |
89.14 | |
CHEBI:271436 | |
89.14 | |
Liquid |
98.5 | |
-70.0°C | |
0.8880g/mL | |
40°C | |
98.5% min. (GC) | |
HOCH2CH2N(CH3)2 | |
MFCD00002846 | |
15, 2845 | |
2-dimethylamino ethanol, n,n-dimethylethanolamine, deanol, dimethylaminoethanol, dimethylethanolamine, norcholine, dmae, dmea, bimanol, liparon | |
UEEJHVSXFDXPFK-UHFFFAOYSA-N | |
2-(dimethylamino)ethanol | |
7902 | |
3.8 mPa.s (20°C) | |
99% | |
N, N-Dimethylethanolamine |
Safety and Handling
GHS H Statement
Toxic if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-542-8
- N,N-Dimethylethanolamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only