Learn More
N,N-Dimethylethylamine, 99%, Thermo Scientific Chemicals
£30.75 - £109.55
Chemical Identifiers
CAS | 598-56-1 |
---|---|
Molecular Formula | C4H11N |
Molecular Weight (g/mol) | 73.13 |
MDL Number | MFCD00009039 |
InChI Key | DAZXVJBJRMWXJP-UHFFFAOYSA-N |
Synonym | n,n-dimethylethylamine, dimethylethylamine, ethanamine, n,n-dimethyl, ethyldimethylamine, n-ethyldimethylamine, ethylamine, n,n-dimethyl, methanamine, n-ethyl-n-methyl, unii-9n5384xvem, c13-15-alkyldimethylamines, amines, c13-15-alkyldimethyl |
PubChem CID | 11723 |
IUPAC Name | N,N-dimethylethanamine |
SMILES | CCN(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10308740
|
Thermo Scientific Acros
219292500 |
250 mL | Glass bottle |
£30.75
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10360844
|
Thermo Scientific
219290010 |
1 L | Glass bottle |
£109.55
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
598-56-1 | |
73.13 | |
DAZXVJBJRMWXJP-UHFFFAOYSA-N | |
11723 | |
CCN(C)C |
C4H11N | |
MFCD00009039 | |
n,n-dimethylethylamine, dimethylethylamine, ethanamine, n,n-dimethyl, ethyldimethylamine, n-ethyldimethylamine, ethylamine, n,n-dimethyl, methanamine, n-ethyl-n-methyl, unii-9n5384xvem, c13-15-alkyldimethylamines, amines, c13-15-alkyldimethyl | |
N,N-dimethylethanamine |
Specifications
598-56-1 | |
100.0 | |
Colorless to Yellow | |
36°C | |
Authentic | |
C4H11N | |
1.3715 to 1.3735 | |
04, 94 | |
n,n-dimethylethylamine, dimethylethylamine, ethanamine, n,n-dimethyl, ethyldimethylamine, n-ethyldimethylamine, ethylamine, n,n-dimethyl, methanamine, n-ethyl-n-methyl, unii-9n5384xvem, c13-15-alkyldimethylamines, amines, c13-15-alkyldimethyl | |
DAZXVJBJRMWXJP-UHFFFAOYSA-N | |
N,N-dimethylethanamine | |
11723 | |
99% | |
N, N-Dimethylethylamine, 99% |
98.5 | |
-140°C | |
0.6750g/mL | |
-36°C | |
99% | |
C2H5N(CH3)2 | |
MFCD00009039 | |
0.675 | |
Solubility in water: soluble. Other solubilities: miscible with most common organic solvents | |
CCN(C)C | |
73.13 | |
73.13 | |
Liquid |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapour.
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 209-940-8
missing translation for 'tsca' : TSCA
- N,N-Dimethylethylamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only