Learn More
N,N'-Dimethylethylenediamine, tech., 85%, Thermo Scientific Chemicals
£49.90 - £141.00
Chemical Identifiers
CAS | 110-70-3 |
---|---|
InChI Key | KVKFRMCSXWQSNT-UHFFFAOYSA-N |
Synonym | n,n'-dimethylethylenediamine, 1,2-bis methylamino ethane, n1,n2-dimethylethane-1,2-diamine, n,n'-dimethyl-1,2-ethanediamine, dimethylethylenediamine, sym-dimethylethylenediamine, 1,2-ethanediamine, n,n'-dimethyl, 2,5-diazahexane, n,n'-dimethyldiaminoethane, n,n'-dimethylethanediamine |
PubChem CID | 8070 |
IUPAC Name | N,N'-dimethylethane-1,2-diamine |
SMILES | CNCCNC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10214240
|
Thermo Scientific Acros
116190250 |
25 g | Glass bottle |
£49.90
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10539420
|
Thermo Scientific Acros
116191000 |
100 g | Glass bottle |
£141.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-70-3 | |
n,n'-dimethylethylenediamine, 1,2-bis methylamino ethane, n1,n2-dimethylethane-1,2-diamine, n,n'-dimethyl-1,2-ethanediamine, dimethylethylenediamine, sym-dimethylethylenediamine, 1,2-ethanediamine, n,n'-dimethyl, 2,5-diazahexane, n,n'-dimethyldiaminoethane, n,n'-dimethylethanediamine | |
N,N'-dimethylethane-1,2-diamine |
KVKFRMCSXWQSNT-UHFFFAOYSA-N | |
8070 | |
CNCCNC |
Specifications
110-70-3 | |
0.8190g/mL | |
26°C | |
C4H12N2 | |
1.4295 | |
0.819 | |
Solubility in water: miscible. Other solubilities: soluble in organic solvents | |
CNCCNC | |
88.15 | |
88.15 | |
Technical | |
N,N′-Dimethylethylenediamine, tech., 85% |
Colorless to Yellow | |
119.0°C | |
Authentic | |
CH3NHCH2CH2NHCH3 | |
04, 250 | |
n,n'-dimethylethylenediamine, 1,2-bis methylamino ethane, n1,n2-dimethylethane-1,2-diamine, n,n'-dimethyl-1,2-ethanediamine, dimethylethylenediamine, sym-dimethylethylenediamine, 1,2-ethanediamine, n,n'-dimethyl, 2,5-diazahexane, n,n'-dimethyldiaminoethane, n,n'-dimethylethanediamine | |
KVKFRMCSXWQSNT-UHFFFAOYSA-N | |
N,N'-dimethylethane-1,2-diamine | |
8070 | |
85% | |
Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-793-3
missing translation for 'rtecsNumber' : KV4250000
missing translation for 'tsca' : TSCA
RUO – Research Use Only