missing translation for 'onlineSavingsMsg'
Learn More
N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals Product Code.: 10157560

N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10157560
25 mL, Glass Bottle
Click to view available options
Quantity:
25 mL
500 mL
1 L
2.5 L
10 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Plastic Drum
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 10157560

Brand: Thermo Scientific Acros 423640250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfaShow MoreShow Less
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Melting Point -61.0°C
CAS Min % 99.8
CAS Max % 100.0
Decomposition Information >350°C
Density 0.9450g/mL
Boiling Point 153.0°C
Quantity 25 mL
Flash Point 58°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4290 to 1.4320
Assay Percent Range 99.8+%
Residue after Evaporation 0.005% max.
Specific Gravity 0.945
Titratable Acid 0.0005 meq/g max.
Linear Formula HCON(CH3)2
Titratable Base 0.003 meq/g max.
Vapor Pressure Vapor Pressure: 4.9mbar at 20°C
Viscosity 0.8 mPa.s (20°C)
Beilstein 04, 58
Water 0.15% Max. (coulometric)
Fieser 01,278; 02,153; 03,115; 04,184; 05,247; 07,124; 08,189; 09,182; 11,198; 12,203; 13,124; 14,148; 16,144
Merck Index 15, 3267
Solubility Information Solubility in water: soluble. Other solubilities: miscible with most organic solvents
Formula Weight 73.09
Percent Purity ≥99.8%
Grade ACS Reagent
Chemical Name or Material N,N-Dimethylformamide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467171 Certificate TypeCertificate of Analysis Date11/11/2024 Catalog Number423645000,423640250,423640010,423640100,423640025
Lot NumberA0427658 Certificate TypeCertificate of Analysis Date31/08/2023 Catalog Number423645000,423640250,423640010,423640100,423640025
Lot NumberA0453633 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number423645000,423640250,423640010,423640100,423640025
Lot NumberA0452226 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number423645000,423640250,423640010,423640100,423640025
Lot NumberA0395297 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number423645000,423640250,423640010,423640100,423640025
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylformamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Reproductive toxicity Category 1B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : N,N-Dimethylformamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.