Learn More
N,N-Dimethylformamide, 99.8%, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals
£26.93 - £380.90
Chemical Identifiers
CAS | 68-12-2 |
---|---|
Molecular Formula | C3H7NO |
Molecular Weight (g/mol) | 73.10 |
MDL Number | MFCD00003284 |
InChI Key | ZMXDDKWLCZADIW-UHFFFAOYSA-N |
Synonym | dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa |
PubChem CID | 6228 |
ChEBI | CHEBI:17741 |
IUPAC Name | N,N-dimethylformamide |
SMILES | CN(C)C=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10094123
|
Thermo Scientific Acros
327171000 |
100 mL |
£26.93
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10009510
|
Thermo Scientific Acros
327175000 |
500 mL |
£44.12
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10617784
|
Thermo Scientific Acros
327170025 |
2.5 L |
£143.72
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10422915
|
Thermo Scientific Acros
327170100 |
10 L |
£380.90
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For molecular biologyChemical Identifiers
68-12-2 | |
73.10 | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
6228 | |
N,N-dimethylformamide |
C3H7NO | |
MFCD00003284 | |
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
CHEBI:17741 | |
CN(C)C=O |
Specifications
-61.0°C | |
68-12-2 | |
100.0 | |
153.0°C | |
Glass Bottle | |
1.4290 to 1.4320 | |
0.001% max. | |
C3H7NO | |
MFCD00003284 | |
0.05% max. (Coulometric) | |
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
N,N-dimethylformamide | |
6228 | |
73.09 | |
Molecular Biology |
(undiluted), (undiluted) at 0nm, 0.1 max. at 280nm, 1 max. at 270nm | |
99.75 | |
0.9450g/mL | |
58°C | |
Liquid | |
99.8% | |
0.945 | |
HCON(CH3)2 | |
0.8 mPa.s (20°C) | |
01,278; 02,153; 03,115; 04,184; 05,247; 07,124; 08,189; 09,182; 11,198; 12,203; 13,124; 14,148; 16,144 | |
Solubility in water: soluble. Other solubilities: miscible with most organic solvents | |
CN(C)C=O | |
73.10 | |
CHEBI:17741 | |
99.8% | |
N,N-Dimethylformamide, DNase, RNase, Protease Free |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May damage the unborn child.
Flammable liquid and vapour.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'einecsNumber' : 200-679-5
missing translation for 'rtecsNumber' : LQ2100000
missing translation for 'tsca' : TSCA
- N,N-Dimethylformamide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Reproductive toxicity Category 1B
Hazard Statement
- H226-Flammable liquid and vapour.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H360D-May damage the unborn child.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : N,N-Dimethylformamide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.