missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylformamide ≥ 99.9%, BAKER ANALYZED™, J.T. Baker™
Click to view available options
1 L
Description
- Suitable as solvent for the analysis of residual solvents of ICH classes 1,2 and 3 according to Ph.Eur. and USP.
- 1 Class solvents residue:
- Benzene: indeterminable
- Carbon tetrachloride: indeterminable
- 1,2-Dichloroethane: indeterminable
- 1,1-Dichloroethane: indeterminable
- 1,1,1-Trichloroethane: indeterminable
- 2 Class solvents residue:
- Acetonitrile: max. 4 ppm
- Cyclohexane: max. 1 ppm
- Dichloromethane: max. 0.6 ppm
- 1,2-Dimethoxyethane: max. 5 ppm
- 1,4-Dioxane: max. 4 ppm
- Hexane: max. 0.3 ppm
- Methanol: max. 1 ppm
- Methylcyclohexane: max. 1 ppm
- Pyridine: max. 1 ppm
- Tetrahydrofuran: max. 0.7 ppm
- Toluene: max. 0.9 ppm
- p-xylen: max. 1 ppm
- m-Xylen: max. 1 ppm
- o-xylene: max. 1 ppm
- 3 Class solvents residue in total: max. 0.5 %
- Acetone: max. 1 ppm
- 1-Butanol: max. 1 ppm
- Butyl acetate: max. 1 ppm
- Ethanol: max. 1 ppm
- Ethyl acetale: max. 1 ppm
- Heptane: max. 1 ppm
- Iso-propyl acetate: max. 1 ppm
- 1-Propanol: max. 1 ppm
- 2-Propanol: max. 1 ppm
Specifications
Specifications
CAS | 68-12-2 |
Acidity | max. 0.0005 meq/g |
Quantity | 1 L |
Refractive Index | Dependent on chemical |
Assay Percent Range | min. 99.9% |
Residue after Evaporation | max. 0.0005% |
Molecular Formula | C3H7NO |
MDL Number | MFCD00003284 |
Water | max. 0.03% |
InChI Key | ZMXDDKWLCZADIW-UHFFFAOYSA-N |
Show More |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Reproductive toxicity Category 1B
- H226-Flammable liquid and vapour.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H360D-May damage the unborn child.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P281-Use personal protective equipment as required.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction