Learn More
N,N-Dimethylformamide, ACS, 99.8+%
Description
N, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Appearance Clear
- Color (APHA) 15
- Evaporation residue 0.005%
- Titratable base 0.003meq/g
- Titratable acid 0.0005meq/g
- H2O 0.15%
Specifications
Specifications
| Melting Point | °C to 61°C |
| Color | Colorless |
| Density | 0.944 g/cm3 at 20°C |
| Boiling Point | 153°C |
| Flash Point | 57°C (135°F) |
| Ignition Point | 440°C (824°F) |
| Assay Percent Range | >99.8% |
| Odor | Amine-like |
| Refractive Index | 1.431 |
| Quantity | 1 L |
| Show More |
Safety and Handling
- N,N-Dimethylformamide
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Reproductive toxicity Category 1B
- H226-Flammable liquid and vapour.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H360D-May damage the unborn child.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : N,N-Dimethylformamide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.