Learn More
N,N-Dimethylformamide-d7, for NMR, 99.5% atom D, Thermo Scientific Chemicals
£69.00 - £800.00
Chemical Identifiers
CAS | 4472-41-7 |
---|---|
Molecular Formula | C3H7NO |
MDL Number | MFCD00003286 |
InChI Key | ZMXDDKWLCZADIW-YYWVXINBSA-N |
Synonym | n,n-dimethylformamide-d7, dmf-d7, deuterated dmf, n,n-di 2h3 methyl 2h formamide, dimethyl formamide-d7, dimethylformamide d, heptadeutero-n,n-dimethylformamide, n,n-dimethylformamide-d7 dmf-d7, formamide-1-d, n,n-di methyl-d3, n,n-dimethylformamide-d7 99.5atom%d |
PubChem CID | 78225 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10479670
|
Thermo Scientific Acros
183600010 |
1 mL | Glass Bottle |
£69.00
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16378186
|
Thermo Scientific Acros
183600050 |
5 mL | Glass Bottle |
£219.00
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10762611
|
Thermo Scientific Acros
183600100 |
10 mL | Glass Bottle |
£395.00
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16388186
|
Thermo Scientific Acros
183600250 |
25 mL | Glass Bottle |
£800.00
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMRChemical Identifiers
4472-41-7 | |
MFCD00003286 | |
n,n-dimethylformamide-d7, dmf-d7, deuterated dmf, n,n-di 2h3 methyl 2h formamide, dimethyl formamide-d7, dimethylformamide d, heptadeutero-n,n-dimethylformamide, n,n-dimethylformamide-d7 dmf-d7, formamide-1-d, n,n-di methyl-d3, n,n-dimethylformamide-d7 99.5atom%d |
C3H7NO | |
ZMXDDKWLCZADIW-YYWVXINBSA-N | |
78225 |
Specifications
Liquid | |
1.0300g/mL | |
58°C | |
0.1% max. (Coulometric) | |
C3H7NO | |
MFCD00003286 | |
ZMXDDKWLCZADIW-YYWVXINBSA-N | |
1-deuterio-N,N-bis(trideuteriomethyl)formamide | |
78225 | |
NMR |
Colorless | |
99.4% min. | |
1.428 | |
4472-41-7 | |
DCON(CD3)2 | |
n,n-dimethylformamide-d7, dmf-d7, deuterated dmf, n,n-di 2h3 methyl 2h formamide, dimethyl formamide-d7, dimethylformamide d, heptadeutero-n,n-dimethylformamide, n,n-dimethylformamide-d7 dmf-d7, formamide-1-d, n,n-di methyl-d3, n,n-dimethylformamide-d7 99.5atom%d | |
CN(C)C=O | |
80.138 | |
80.15 | |
N, N-Dimethylformamide-d7, 99.5 atom % D |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May damage the unborn child.
Flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a gu
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 224-745-8
- N,N-Dimethylformamide-d7
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1A
Hazard Statement
- H226-Flammable liquid and vapour.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H319-Causes serious eye irritation.
- H360D-May damage the unborn child.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only