missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylformamide di-n-propyl acetal, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
N,N-Dimethylformamide di-n-propyl acetal, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N,N-Dimethylformamide di-n-propyl acetal, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
It hydrolyzes with water.
Notes
Moisture, water Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
N,N-Dimethylformamide di-n-propyl acetal, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
It hydrolyzes with water.
Notes
Moisture, water Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 0.854 |
Boiling Point | 67°C to 68°C (14 mmHg) |
Flash Point | 46°C (115°F) |
Linear Formula | (CH3)2NCH(OCH2CH2CH3)2 |
Refractive Index | 1.4088 |
Quantity | 10 g |
UN Number | UN1993 |
Beilstein | 1098524 |
Sensitivity | Moisture sensitive |
Solubility Information | It hydrolyzes with water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'