missing translation for 'onlineSavingsMsg'
Learn More
N,N-Dimethylformamide di-tert-butyl Acetal, tech. 90%, Thermo Scientific Chemicals Product Code.: 11390034

N,N-Dimethylformamide di-tert-butyl Acetal, tech. 90%, Thermo Scientific Chemicals

Product Code. 11390034
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11390034

Brand: Thermo Scientific Alfa Aesar L00431.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N,N-Dimethylformamide di-tert-butyl acetal is used as a reagent in the synthesis of (S)-2-(1-tert-butoxycarbonyl-2-{4-[2-(5-methyl-2-phenyloxazol-4-yl)ethoxy]phenyl}ethylamino)benzoic acid methyl ester, 3-O-tert-butylmorphine, tert-butylesters of pyrrole and indolecarboxylic acids. Further, it is used in the preparation of indolizines by intermolecular cyclization of picolinium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylformamide di-tert-butyl acetal is used as a reagent in the synthesis of (S)-2-(1-tert-butoxycarbonyl-2-{4-[2-(5-methyl-2-phenyloxazol-4-yl)ethoxy]phenyl}ethylamino)benzoic acid methyl ester, 3-O-tert-butylmorphine, tert-butylesters of pyrrole and indolecarboxylic acids. Further, it is used in the preparation of indolizines by intermolecular cyclization of picolinium salts.

Solubility
Miscible with toluene and benzene.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36805-97-7
Molecular Formula C11H25NO2
Molecular Weight (g/mol) 203.326
MDL Number MFCD00015002
InChI Key DBNQIOANXZVWIP-UHFFFAOYSA-N
Synonym n,n-dimethylformamide di-tert-butyl acetal, 1,1-di-tert-butoxytrimethylamine, 1,1-di-tert-butoxy-n,n-dimethylmethanamine, bis tert-butoxy methyl dimethylamine, 1,1-bis 1,1-dimethylethoxy-n,n,n-trimethylamine, di-tert-butoxymethyldimethylamine, n,n-dimethylformamide di-t-butyl acetal, methanamine, 1,1-bis 1,1-dimethylethoxy-n,n-dimethyl, 1,1-ditert-butoxy-n,n-dimethylmethanamine, 1,1-di-tert-butoxy-n,n-dimethylmethylamine
PubChem CID 547712
IUPAC Name N,N-dimethyl-1,1-bis[(2-methylpropan-2-yl)oxy]methanamine
SMILES CC(C)(C)OC(N(C)C)OC(C)(C)C

Specifications

Density 0.85
Boiling Point 75°C to 77°C (12 mmHg)
Flash Point 33°C (91°F)
Linear Formula (CH3)2NCH[OC(CH3)3]2
Refractive Index 1.412
Quantity 25 g
UN Number 1993
Beilstein 969629
Sensitivity Moisture sensitive
Solubility Information Miscible with toluene and benzene.
Formula Weight 203.33
Percent Purity 90%
Grade Technical
Chemical Name or Material N,N-Dimethylformamide di-tert-butyl Acetal
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10244773 Certificate TypeCertificate of Analysis Date24/11/2022 Catalog NumberL00431.06,L00431.14
Lot NumberH31Q20 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL00431.06,L00431.14
Lot Number10189789 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL00431.06,L00431.14
Lot Number10210245 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL00431.06,L00431.14
Lot NumberI29Q028 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL00431.06,L00431.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N-Dimethylformamide di-tert-butyl Acetal, tech. 90%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.