missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylformamide dibenzyl acetal, tech. 80%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
N,N-Dimethylformamide diethyl acetal was used in quantification of cocaine and its primary metabolite, benzoyl ecgonine from urine matrix by gas chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N,N-Dimethylformamide diethyl acetal was used in quantification of cocaine and its primary metabolite, benzoyl ecgonine from urine matrix by gas chromatography.
Solubility
Soluble in variety of inert solvents.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
N,N-Dimethylformamide diethyl acetal was used in quantification of cocaine and its primary metabolite, benzoyl ecgonine from urine matrix by gas chromatography.
Solubility
Soluble in variety of inert solvents.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.045 |
Boiling Point | 138°C to 140°C (0.5 mmHg) |
Quantity | 50 g |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in variety of inert solvents. |
Formula Weight | 271.36 |
Percent Purity | 85% |
Grade | Technical |
Chemical Name or Material | N,N-Dimethylformamide dibenzyl acetal |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'