missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylformamide diethyl acetal, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Density | 0.859 |
Boiling Point | 134°C to 136°C |
Flash Point | 17°C (62°F) |
Refractive Index | 1.399 |
Quantity | 50 g |
UN Number | UN1993 |
Beilstein | 741889 |
Sensitivity | Moisture sensitive |
Formula Weight | 147.22 |
Percent Purity | 95% |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
5004Q18S | Certificate of Analysis | 26/01/2025 | L00205.18 |
10251614 | Certificate of Analysis | 30/08/2024 | L00205.18,L00205.09 |
10243444 | Certificate of Analysis | 14/09/2022 | L00205.18,L00205.09 |
10232440 | Certificate of Analysis | 08/12/2021 | L00205.18,L00205.09 |
J28Q016 | Certificate of Analysis | 08/12/2021 | L00205.18,L00205.09 |
Safety and Handling


Signal Word
- Warning
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : N,N-Dimethylformamide diethyl acetal
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
N,N-Dimethylformamide diethyl acetal, 95%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction