missing translation for 'onlineSavingsMsg'
Learn More
N,N-Dimethylformamide diethyl acetal, 95%, Thermo Scientific Chemicals Product Code.: 10478710

N,N-Dimethylformamide diethyl acetal, 95%, Thermo Scientific Chemicals

Product Code. 10478710
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 10478710

Brand: Thermo Scientific Acros 162080100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1188-33-6
Molecular Formula C7H17NO2
Molecular Weight (g/mol) 147.22
MDL Number MFCD00009227
InChI Key BWKAYBPLDRWMCJ-UHFFFAOYSA-N
Synonym n,n-dimethylformamide diethyl acetal, 1,1-diethoxytrimethylamine, methanamine, 1,1-diethoxy-n,n-dimethyl, n,n-dimethyformamide diethy acetal, diethoxymethyl dimethylamine, n,n-dimethylformamide diethylacetal, dimethylformamide diethylacetal, 1,1-diethoxy trimethyl amine, 1,1-diethoxy-n,n-dimethylmethylamine, 1,1-diethoxy-n,n-dimethyl-methanamine
PubChem CID 70913
IUPAC Name 1,1-diethoxy-N,N-dimethylmethanamine
SMILES CCOC(N(C)C)OCC

Specifications

CAS Min % 94.0
CAS Max % 100.0
Density 0.8590g/mL
Boiling Point 128°C to 130°C
Flash Point 28°C
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Linear Formula (CH3)2NCH(OC2H5)2
Quantity 10 g
Fieser 01,281; 02,154; 05,253; 07,125; 10,157
Specific Gravity 0.859
Solubility Information Solubility in water: decomposes
Formula Weight 147.22
Percent Purity 95%
Chemical Name or Material N,N-Dimethylformamide diethyl acetal
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468463 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number162080100,162080500
Lot NumberA0467237 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog Number162080100,162080500
Lot NumberA0464715 Certificate TypeCertificate of Analysis Date22/07/2024 Catalog Number162080100,162080500
Lot NumberA0431197 Certificate TypeCertificate of Analysis Date11/12/2023 Catalog Number162080100,162080500
Lot NumberA0393601 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number162080100,162080500
compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylformamide diethyl acetal
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N-Dimethylformamide diethyl acetal, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.