missing translation for 'onlineSavingsMsg'
Learn More
N,N-Dimethylformamide dimethyl acetal, 94%, Thermo Scientific Chemicals Product Code.: 11428276

N,N-Dimethylformamide dimethyl acetal, 94%, Thermo Scientific Chemicals

Product Code. 11428276
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 11428276

Brand: Thermo Scientific Alfa Aesar A15350.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

N,N-Dimethylformamide dimethyl acetal is used as an intermediate in the formation of pyridine derivatives, which exhibits inhibition against PI3 kinase p110alfa enzymes. It is utilized for the derivatization of primary sulfonamides and trifluoroacetic acid. It is also used in the preparation of formamidine derivatives. It is used as a reagent for n-dimethylaminomethylene and methyl esters. Further, it is used to catalyze the coupling of epoxides with carbon dioxide to prepare cyclic carbonates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylformamide dimethyl acetal is used as an intermediate in the formation of pyridine derivatives, which exhibits inhibition against PI3 kinase p110alfa enzymes. It is utilized for the derivatization of primary sulfonamides and trifluoroacetic acid. It is also used in the preparation of formamidine derivatives. It is used as a reagent for n-dimethylaminomethylene and methyl esters. Further, it is used to catalyze the coupling of epoxides with carbon dioxide to prepare cyclic carbonates.

Solubility
Miscible with most organic solvents.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4637-24-5
Molecular Formula C5H13NO2
Molecular Weight (g/mol) 119.16
MDL Number MFCD00008482
InChI Key ZSXGLVDWWRXATF-UHFFFAOYSA-N
Synonym n,n-dimethylformamide dimethyl acetal, 1,1-dimethoxytrimethylamine, dimethoxymethyl dimethylamine, methanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylformamide-dimethylacetal, dmf dimethyl acetal, dmfdma, dimethylformamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylmethylamine, dimethoxy dimethylamino methaneShow MoreShow Less
PubChem CID 78373
ChEBI CHEBI:85061
IUPAC Name (dimethoxymethyl)dimethylamine
SMILES COC(OC)N(C)C

Specifications

Melting Point -85°C
Density 0.897
Boiling Point 102°C to 104°C
Flash Point 7°C (45°F)
Refractive Index 1.3972
Quantity 100 g
UN Number UN1993
Sensitivity Moisture sensitive
Solubility Information Miscible with most organic solvents.
Formula Weight 119.16
Percent Purity 97%
Chemical Name or Material N,N-Dimethylformamide dimethyl acetal
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10239341 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA15350.14,A15350.36,A15350.22
Lot Number10166144 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15350.14,A15350.36,A15350.22
Lot NumberX29A061 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15350.14,A15350.36,A15350.22
Lot NumberH25R011 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15350.14,A15350.36,A15350.22
Lot Number10145851 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15350.14,A15350.36,A15350.22
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylformamide dimethyl acetal
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : N,N-Dimethylformamide dimethyl acetal

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N-Dimethylformamide dimethyl acetal, 94%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.