missing translation for 'onlineSavingsMsg'
Learn More
N,N-Dimethylformamide dimethyl acetal, ca. 97%, AcroSeal™, Thermo Scientific Chemicals Product Code.: 11905301

N,N-Dimethylformamide dimethyl acetal, ca. 97%, AcroSeal™, Thermo Scientific Chemicals

Product Code. 11905301
100 mL, AcroSeal™ Glass bottle
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 11905301

Brand: Thermo Scientific Acros 446151000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 4637-24-5, 149-73-5, 68-12-2, 67-56-1
Color White
Density 0.8900g/mL
Boiling Point 102.0°C to 104.0°C
Flash Point 7°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging AcroSeal™ Glass bottle
Molecular Formula C5H13NO2
Linear Formula (CH3)2NCH(OCH3)2
Refractive Index 1.3972
MDL Number MFCD00008482
Quantity 100 mL
Specific Gravity 0.89
Synonym n,n-dimethylformamide dimethyl acetal, 1,1-dimethoxytrimethylamine, dimethoxymethyl dimethylamine, methanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylformamide-dimethylacetal, dmf dimethyl acetal, dmfdma, dimethylformamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylmethylamine, dimethoxy dimethylamino methane
Solubility Information Solubility in water: hydrolysis. Other solubilities: soluble in many organic solvents
InChI Key ZSXGLVDWWRXATF-UHFFFAOYSA-N
SMILES COC(OC)N(C)C
IUPAC Name (dimethoxymethyl)dimethylamine
Molecular Weight (g/mol) 119.16
PubChem CID 78373
ChEBI CHEBI:85061
Formula Weight 119.16
Percent Purity ca. 97%
Physical Form Solid
Chemical Name or Material N, N-Dimethylformamide dimethyl acetal
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylformamide dimethyl acetal
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • Reproductive toxicity Category 1B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N-Dimethylformamide dimethyl acetal, ca. 97%, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.