missing translation for 'onlineSavingsMsg'
Learn More
N,N-Dimethylformamide dineopentyl acetal, 98%, Thermo Scientific Chemicals Product Code.: 11397805

N,N-Dimethylformamide dineopentyl acetal, 98%, Thermo Scientific Chemicals

Product Code. 11397805
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11397805

Brand: Thermo Scientific Alfa Aesar L02728.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N,N-Dimethylformamide dineopentyl acetal helps in the lactonization of ω-hydroxyacids, giving lactones of up to 16-membered rings. It can be used to convert primary alcohols to alkylating agents for use in the alkylation of thiols. It is used in the esterification of Nα-9-fluorenylmethyloxycarbonylamino acids. It was used in the synthesis of 1,3-dialkyl, benzyl and cyclohexyl barbiturate derivatives. It was used as reagent during the synthesis of L-serine and L-cystine stereospecifically labeled with deuterium at the β-position.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylformamide dineopentyl acetal helps in the lactonization of ω-hydroxyacids, giving lactones of up to 16-membered rings. It can be used to convert primary alcohols to alkylating agents for use in the alkylation of thiols. It is used in the esterification of Nα-9-fluorenylmethyloxycarbonylamino acids. It was used in the synthesis of 1,3-dialkyl, benzyl and cyclohexyl barbiturate derivatives. It was used as reagent during the synthesis of L-serine and L-cystine stereospecifically labeled with deuterium at the β-position.

Solubility
Hydrolyzes with water.

Notes
Moisture Sensitive. Incompatible with water and oxidizing agents. Store under dry inert gas. Protect from humidity and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4909-78-8
Molecular Formula C13H30NO2
Molecular Weight (g/mol) 232.39
MDL Number MFCD00008851
InChI Key KEXFRBIOHPDZQM-UHFFFAOYSA-O
Synonym n,n-dimethylformamide dineopentyl acetal, methanamine, 1,1-bis 2,2-dimethylpropoxy-n,n-dimethyl, n,n-dimethyl-1,1-bis neopentyloxy methanamine, dimethylformamide dineopentyl acetal, 1,1-bis 2,2-dimethylpropoxy-n,n-dimethylmethanamine, bis 2,2-dimethylpropoxy methyl dimethylamine, acmc-1al8s, 1,1-bis 2,2-dimethylpropoxy-n,n,n-trimethylamine, 1,1-dineopentyloxytrimethylamine, dimethylformamide dineopentylacetalShow MoreShow Less
PubChem CID 78623
IUPAC Name 1,1-bis(2,2-dimethylpropoxy)-N,N-dimethylmethanamine
SMILES C[NH+](C)C(OCC(C)(C)C)OCC(C)(C)C

Specifications

Density 0.83
Boiling Point 85°C to 87°C (10 mmHg)
Flash Point 63°C (145°F)
Linear Formula (CH3)2NCH[OCH2C(CH3)3]2
Refractive Index 1.4117
Quantity 50 g
Beilstein 741992
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes with water.
Formula Weight 231.38
Percent Purity 98%
Chemical Name or Material N,N-Dimethylformamide dineopentyl acetal
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10243501 Certificate TypeCertificate of Analysis Date15/09/2022 Catalog NumberL02728.18,L02728.09
Lot Number10181578 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL02728.18,L02728.09
Lot Number10229294 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL02728.18,L02728.09
Lot Number10181577 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL02728.18,L02728.09
Lot Number10149274 Certificate TypeCertificate of Analysis Date02/12/2021 Catalog NumberL02728.18,L02728.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N-Dimethylformamide dineopentyl acetal, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.