missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, puriss. p.a. ACS Reagent, Honeywell

£74.52 - £2090.11

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15643900
View Documents Promotion Details
Honeywell Riedel-de Haën
33120-1L
1 L
£74.52
1 L
Please to purchase this item. Need a web account? Register with us today!
15653900
View Documents
Honeywell Riedel-de Haën
33120-2.5L
2.5 L
£147.52
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15673900
View Documents
Honeywell Riedel-de Haën
33120-25L
25 L
£885.42
25 L
Please to purchase this item. Need a web account? Register with us today!
15633900
View Documents
Honeywell Riedel-de Haën
33120-180KG
180 kg
£2090.11
180kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

68-12-2
73.10
ZMXDDKWLCZADIW-UHFFFAOYSA-N
6228
N,N-dimethylformamide
C3H7NO
MFCD00003284
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
CHEBI:17741
CN(C)C=O

Specifications

68-12-2
0.152
833°F
HCON(CH3)2
2.7mmHg (20°C)
605365
ZMXDDKWLCZADIW-UHFFFAOYSA-N
N,N-dimethylformamide
6228
73.09g/mol
ACS/puriss. p.a.
0.944g/mL, 0.948 to 0.950g/mL at 20°C
58°C (136.4°F)
C3H7NO
MFCD00003284
2265
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
CN(C)C=O
73.10
CHEBI:17741
≥99.8% (GC)
N,N-Dimethylformamide, Reag. Ph. Eur.
Videos
Safety and Handling

Safety and Handling

P201-P210-P261-P280-P308 + P313-P370 + P378

H226-H312 + H332-H319-H360D

Danger

missing translation for 'einecsNumber' : 200-679-5

missing translation for 'rtecsNumber' : LQ2100000

Product Identifier
  • N,N-Dimethylformamide

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H360D-May damage the unborn child.
  • H332-Harmful if inhaled.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : N,N-dimethylformamide

SDS
Documents

Documents

Product Certifications