missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals N,N-Dimethylglycine hydrochloride, 99%
£27.90 - £296.00
Chemical Identifiers
CAS | 2491-06-7 |
---|---|
Molecular Formula | C4H9NO2·HCl |
Molecular Weight (g/mol) | 139.58 |
MDL Number | MFCD00012610 |
InChI Key | FKASAVXZZLJTNX-UHFFFAOYSA-N |
Synonym | n,n-dimethylglycine hydrochloride, dimethylglycine hydrochloride, glycine, n,n-dimethyl-, hydrochloride, n-methylsarcosine hydrochloride, n,n-dimethylglycine hcl, dimethylamino acetic acid hydrochloride, unii-yxk75eae92, yxk75eae92, n,n-dimethylaminoacetic acid hydrochloride, 2-dimethylamino acetic acid hydrochloride |
PubChem CID | 75605 |
IUPAC Name | 2-(dimethylamino)acetic acid;hydrochloride |
SMILES | CN(C)CC(=O)O.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10431441
|
Thermo Scientific Chemicals
204660050 |
5 g | Glass bottle |
£27.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10378290
|
Thermo Scientific Chemicals
204660250 |
25 g | Glass bottle |
£97.20
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10026293
|
Thermo Scientific Chemicals
204661000 |
100 g | Glass bottle |
£296.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2491-06-7 | |
139.58 | |
FKASAVXZZLJTNX-UHFFFAOYSA-N | |
75605 | |
CN(C)CC(=O)O.Cl |
C4H9NO2·HCl | |
MFCD00012610 | |
n,n-dimethylglycine hydrochloride, dimethylglycine hydrochloride, glycine, n,n-dimethyl-, hydrochloride, n-methylsarcosine hydrochloride, n,n-dimethylglycine hcl, dimethylamino acetic acid hydrochloride, unii-yxk75eae92, yxk75eae92, n,n-dimethylaminoacetic acid hydrochloride, 2-dimethylamino acetic acid hydrochloride | |
2-(dimethylamino)acetic acid;hydrochloride |
Specifications
188°C to 191°C | |
White | |
100.0 | |
99% | |
(CH3)2NCH2CO2H·HCl | |
04, I, 469 | |
n,n-dimethylglycine hydrochloride, dimethylglycine hydrochloride, glycine, n,n-dimethyl-, hydrochloride, n-methylsarcosine hydrochloride, n,n-dimethylglycine hcl, dimethylamino acetic acid hydrochloride, unii-yxk75eae92, yxk75eae92, n,n-dimethylaminoacetic acid hydrochloride, 2-dimethylamino acetic acid hydrochloride | |
FKASAVXZZLJTNX-UHFFFAOYSA-N | |
2-(dimethylamino)acetic acid;hydrochloride | |
75605 | |
99% | |
N, N-Dimethylglycine hydrochloride |
2491-06-7 | |
98.5 | |
Crystalline Powder | |
C4H9NO2·HCl | |
MFCD00012610 | |
15, 3269 | |
Solubility in water: 100g/L (20 c). Other solubilities: insoluble in chloroform and acetone | |
CN(C)CC(=O)O.Cl | |
139.58 | |
139.58 | |
Authentic |
Safety and Handling
missing translation for 'einecsNumber' : 219-648-2
RUO – Research Use Only