Learn More
N,N-Dimethylguanidine sulfate, 98%, Thermo Scientific Chemicals
£40.80 - £267.00
Chemical Identifiers
CAS | 598-65-2 |
---|---|
Molecular Formula | 0·5 H2SO4 |
Molecular Weight (g/mol) | 272.32 |
MDL Number | MFCD00013131 |
InChI Key | QSCHFHVDZCPIKX-UHFFFAOYSA-N |
Synonym | 1,1-dimethylguanidine sulfate 2:1, guanidine, n,n-dimethyl-, sulfate 2:1, bis dimethylguanidine ; sulfuric acid, 1,1-dimethylguanidine hemisulfate, pubchem19198, guanidine,n,n-dimethyl, 1,1-dimethyl guanidinium sulfate, 1,1-dimethylguanidine sulfate salt, 1,1-dimethylguanidinium sulphate 2:1 |
PubChem CID | 69024 |
IUPAC Name | 1,1-dimethylguanidine;sulfuric acid |
SMILES | CN(C)C(=N)N.CN(C)C(=N)N.OS(=O)(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10730565
|
Thermo Scientific Acros
408350100 |
10 g | Plastic bottle |
£40.80
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10657941
|
Thermo Scientific Acros
408350500 |
50 g | Plastic bottle |
£143.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10442971
|
Thermo Scientific Acros
408351000 |
100 g | Plastic bottle |
£267.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
598-65-2 | |
272.32 | |
QSCHFHVDZCPIKX-UHFFFAOYSA-N | |
69024 | |
CN(C)C(=N)N.CN(C)C(=N)N.OS(=O)(=O)O |
0·5 H2SO4 | |
MFCD00013131 | |
1,1-dimethylguanidine sulfate 2:1, guanidine, n,n-dimethyl-, sulfate 2:1, bis dimethylguanidine ; sulfuric acid, 1,1-dimethylguanidine hemisulfate, pubchem19198, guanidine,n,n-dimethyl, 1,1-dimethyl guanidinium sulfate, 1,1-dimethylguanidine sulfate salt, 1,1-dimethylguanidinium sulphate 2:1 | |
1,1-dimethylguanidine;sulfuric acid |
Specifications
598-65-2 | |
100.0 | |
White | |
98% | |
(CH)2NC(NH)NH2)2·H2SO4 | |
04,II,574 | |
Solubility in water: 490g/L (20°C) | |
CN(C)C(=N)N.CN(C)C(=N)N.OS(=O)(=O)O | |
272.32 | |
272.32 | |
Powder |
97.5 | |
280.0°C | |
Authentic | |
0·5 H2SO4 | |
MFCD00013131 | |
1,1-dimethylguanidine sulfate 2:1, guanidine, n,n-dimethyl-, sulfate 2:1, bis dimethylguanidine ; sulfuric acid, 1,1-dimethylguanidine hemisulfate, pubchem19198, guanidine,n,n-dimethyl, 1,1-dimethyl guanidinium sulfate, 1,1-dimethylguanidine sulfate salt, 1,1-dimethylguanidinium sulphate 2:1 | |
QSCHFHVDZCPIKX-UHFFFAOYSA-N | |
1,1-dimethylguanidine;sulfuric acid | |
69024 | |
98% | |
N, N-Dimethylguanidine sulfate, 93% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
missing translation for 'einecsNumber' : 209-946-0
missing translation for 'tsca' : TSCA
RUO – Research Use Only