Learn More
N,N-Dimethylhexylamine, 99%, Thermo Scientific Chemicals
£37.89 - £392.96
Chemical Identifiers
CAS | 4385-04-0 |
---|---|
Molecular Formula | C8H19N |
Molecular Weight (g/mol) | 129.247 |
MDL Number | MFCD00009522 |
InChI Key | QMHNQZGXPNCMCO-UHFFFAOYSA-N |
Synonym | n,n-dimethylhexylamine, hexyldimethylamine, 1-hexanamine, n,n-dimethyl, 1-dimethylaminohexane, n,n-dimethyl-n-hexylamine, dimethylhexylamine, hexyl-dimethyl-amine, n-hexyl dimethylamine, hexanamine, n,n-dimethyl, acmc-1ct1t |
PubChem CID | 78090 |
IUPAC Name | N,N-dimethylhexan-1-amine |
SMILES | CCCCCCN(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11413607
|
Thermo Scientific Alfa Aesar
A17364.06 |
5 g |
£37.89
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11423607
|
Thermo Scientific Alfa Aesar
A17364.14 |
25 g |
£124.62
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11433607
|
Thermo Scientific Alfa Aesar
A17364.22 |
100 g |
£392.96
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N,N-Dimethylhexylamine is used as an ion-pairing agent to study the poor retention of uridine diphosphate-linked intermediates on reverse phase media. It is also used in the determination of five monophosphate nucleotides in baby foods. It acts as an additive and encapsulated into the nanoparticle with salinomycin (sali), which has selective toxicity to cancer stem cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N-Dimethylhexylamine is used as an ion-pairing agent to study the poor retention of uridine diphosphate-linked intermediates on reverse phase media. It is also used in the determination of five monophosphate nucleotides in baby foods. It acts as an additive and encapsulated into the nanoparticle with salinomycin (sali), which has selective toxicity to cancer stem cells.
Solubility
Miscible with methanol.
Notes
Incompatible with strong acids and strong oxidizing agents.
Chemical Identifiers
4385-04-0 | |
129.247 | |
QMHNQZGXPNCMCO-UHFFFAOYSA-N | |
78090 | |
CCCCCCN(C)C |
C8H19N | |
MFCD00009522 | |
n,n-dimethylhexylamine, hexyldimethylamine, 1-hexanamine, n,n-dimethyl, 1-dimethylaminohexane, n,n-dimethyl-n-hexylamine, dimethylhexylamine, hexyl-dimethyl-amine, n-hexyl dimethylamine, hexanamine, n,n-dimethyl, acmc-1ct1t | |
N,N-dimethylhexan-1-amine |
Specifications
4385-04-0 | |
146°C to 150°C | |
C8H19N | |
MFCD00009522 | |
n,n-dimethylhexylamine, hexyldimethylamine, 1-hexanamine, n,n-dimethyl, 1-dimethylaminohexane, n,n-dimethyl-n-hexylamine, dimethylhexylamine, hexyl-dimethyl-amine, n-hexyl dimethylamine, hexanamine, n,n-dimethyl, acmc-1ct1t | |
QMHNQZGXPNCMCO-UHFFFAOYSA-N | |
N,N-dimethylhexan-1-amine | |
78090 | |
99% |
0.744 | |
33°C (92°F) | |
1.4136 | |
UN2733 | |
Miscible with methanol. | |
CCCCCCN(C)C | |
129.247 | |
129.25 | |
N,N-Dimethylhexylamine |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 224-491-8
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.