missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylmethyleneiminium iodide, 97%

Product Code. 10461051 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 10461051

Brand: Thermo Scientific Acros 201010500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 33797-51-2 | C3H8IN | 185.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33797-51-2
Molecular Formula C3H8IN
Molecular Weight (g/mol) 185.01
MDL Number MFCD00011810
InChI Key VVDUZZGYBOWDSQ-UHFFFAOYSA-M
Synonym eschenmoser's salt, n,n-dimethylmethyleneiminium iodide, n-methyl-n-methylenemethanaminium iodide, dimethyl methylidene azanium iodide, n,n-dimethyl methyleneammonium iodide, dimethyl methylene ammonium iodide, methanaminium, n-methyl-n-methylene-, iodide, eschenmoser inverted exclamation mark s salt, eschenmoser salt, dimethyl methylene ammounium iodide
PubChem CID 2724133
IUPAC Name dimethyl(methylidene)azanium;iodide
SMILES C[N+](=C)C.[I-]

Specifications

Melting Point 219°C
Color Cream to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula H2C=N(CH3)2I
Quantity 50 g
Beilstein 04, IV, 153
Fieser 04,186; 08,194; 10,160; 13,121
Merck Index 15, 3275
Formula Weight 185.01
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material N, N-Dimethylmethyleneiminium iodide, 97%
Show More Show Less
compliance-icons
Product Identifier
  • N,N-Dimethylmethyleneiminium iodide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.