Learn More
N,N-Dimethyloctylamine, 97%, Thermo Scientific Chemicals
£37.60 - £1560.00
Chemical Identifiers
CAS | 7378-99-6 |
---|---|
Molecular Formula | C10H23N |
Molecular Weight (g/mol) | 157.30 |
MDL Number | MFCD00009558 |
InChI Key | UQKAOOAFEFCDGT-UHFFFAOYSA-N |
Synonym | n,n-dimethyloctylamine, n,n-dimethyl-n-octylamine, dimethyloctylamine, octyldimethylamine, dimethyl octyl amine, n-octyldimethylamine, dimethyl-n-octylamine, 1-octanamine, n,n-dimethyl, octylamine, n,n-dimethyl, unii-20n7h7x4sd |
PubChem CID | 16224 |
IUPAC Name | N,N-dimethyloctan-1-amine |
SMILES | CCCCCCCCN(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10206570
|
Thermo Scientific Acros
225290250 |
25 mL | Glass bottle |
£37.60
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10657552
|
Thermo Scientific Acros
225295000 |
500 mL | Glass bottle |
£472.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10105163
|
Thermo Scientific Acros
225290025 |
2.5 L | Glass bottle |
£1560.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7378-99-6 | |
157.30 | |
UQKAOOAFEFCDGT-UHFFFAOYSA-N | |
16224 | |
CCCCCCCCN(C)C |
C10H23N | |
MFCD00009558 | |
n,n-dimethyloctylamine, n,n-dimethyl-n-octylamine, dimethyloctylamine, octyldimethylamine, dimethyl octyl amine, n-octyldimethylamine, dimethyl-n-octylamine, 1-octanamine, n,n-dimethyl, octylamine, n,n-dimethyl, unii-20n7h7x4sd | |
N,N-dimethyloctan-1-amine |
Specifications
7378-99-6 | |
100.0 | |
0.7650g/mL | |
65°C | |
96% min. (C8-chains) (GC) | |
CH3(CH2)7N(CH3)2 | |
MFCD00009558 | |
0.765 | |
Solubility in water: slightly soluble | |
CCCCCCCCN(C)C | |
157.30 | |
157.3 | |
Liquid |
96.0 | |
-57.0°C | |
195.0°C | |
Authentic | |
C10H23N | |
1.4233 to 1.4253 | |
04,I,386 | |
n,n-dimethyloctylamine, n,n-dimethyl-n-octylamine, dimethyloctylamine, octyldimethylamine, dimethyl octyl amine, n-octyldimethylamine, dimethyl-n-octylamine, 1-octanamine, n,n-dimethyl, octylamine, n,n-dimethyl, unii-20n7h7x4sd | |
UQKAOOAFEFCDGT-UHFFFAOYSA-N | |
N,N-dimethyloctan-1-amine | |
16224 | |
97% | |
N, N-Dimethyloctylamine, 97% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
Very toxic to aquatic life.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contac
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 230-939-3
missing translation for 'rtecsNumber' : RG8075000
missing translation for 'tsca' : TSCA
- N,N-Dimethyloctylamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H301-Toxic if swallowed.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P273-Avoid release to the environment.
RUO – Research Use Only