missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N,N',N'-Tetramethyl-p-phenylenediamine, 98%, Thermo Scientific Chemicals
£29.20 - £333.00
Chemical Identifiers
CAS | 100-22-1 |
---|---|
Molecular Formula | C10H16N2 |
Molecular Weight (g/mol) | 164.25 |
InChI Key | CJAOGUFAAWZWNI-UHFFFAOYSA-N |
Synonym | wurster's blue, wurster's reagent, n,n,n',n'-tetramethyl-p-phenylenediamine, n,n,n',n'-tetramethyl-1,4-phenylenediamine, tetramethyl-p-phenylenediamine, 1,4-bis dimethylamino benzene, p-bis dimethylamino benzene, 1,4-benzenediamine, n,n,n',n'-tetramethyl, benzene, 1,4-bis dimethylamino, unii-p4p3ac32zb |
PubChem CID | 7490 |
IUPAC Name | 1-N,1-N,4-N,4-N-tetramethylbenzene-1,4-diamine |
SMILES | CN(C)C1=CC=C(C=C1)N(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10327640
|
Thermo Scientific Acros
170790050 |
5 g | Glass bottle |
£29.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10215990
|
Thermo Scientific Acros
170790250 |
25 g | Glass bottle |
£94.10
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11995261
|
Thermo Scientific Acros
170791000 |
100 g | Glass Bottle |
£333.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
100-22-1 | |
164.25 | |
wurster's blue, wurster's reagent, n,n,n',n'-tetramethyl-p-phenylenediamine, n,n,n',n'-tetramethyl-1,4-phenylenediamine, tetramethyl-p-phenylenediamine, 1,4-bis dimethylamino benzene, p-bis dimethylamino benzene, 1,4-benzenediamine, n,n,n',n'-tetramethyl, benzene, 1,4-bis dimethylamino, unii-p4p3ac32zb | |
1-N,1-N,4-N,4-N-tetramethylbenzene-1,4-diamine |
C10H16N2 | |
CJAOGUFAAWZWNI-UHFFFAOYSA-N | |
7490 | |
CN(C)C1=CC=C(C=C1)N(C)C |
Specifications
100-22-1 | |
Beige-Brown to Brown | |
>112°C | |
97.5% min. (GC) | |
C6H4[N(CH3)2]2 | |
15,9376 | |
Solubility in water: slightly in cold, more in hot water. Other solubilities: soluble in alcohols, chloroform, ether | |
CN(C)C1=CC=C(C=C1)N(C)C | |
164.25 | |
164.25 | |
Crystalline Flakes |
48.0°C to 52.0°C | |
260.0°C | |
Authentic | |
C10H16N2 | |
13,74 | |
wurster's blue, wurster's reagent, n,n,n',n'-tetramethyl-p-phenylenediamine, n,n,n',n'-tetramethyl-1,4-phenylenediamine, tetramethyl-p-phenylenediamine, 1,4-bis dimethylamino benzene, p-bis dimethylamino benzene, 1,4-benzenediamine, n,n,n',n'-tetramethyl, benzene, 1,4-bis dimethylamino, unii-p4p3ac32zb | |
CJAOGUFAAWZWNI-UHFFFAOYSA-N | |
1-N,1-N,4-N,4-N-tetramethylbenzene-1,4-diamine | |
7490 | |
98% | |
N,N,N',N'-Tetramethyl-p-phenylenediamine |
Safety and Handling
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 202-831-6
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- N,N,N',N'-Tetramethyl-p-phenylenediamine
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only