missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride, 98+%

Product Code. 10572764 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10572764

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 637-01-4 | C10H16N2·2HCl | 237.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-01-4
Molecular Formula C10H16N2·2HCl
Molecular Weight (g/mol) 237.17
MDL Number MFCD00012482
InChI Key FBHKTSXMTASXFJ-UHFFFAOYSA-N
Synonym n,n,n',n'-tetramethyl-p-phenylenediamine dihydrochloride, wurster's reagent dihydrochloride, n,n,n',n'-tetramethyl-1,4-phenylenediamine dihydrochloride, unii-66w8hka51x, n1,n1,n4,n4-tetramethylbenzene-1,4-diamine dihydrochloride, wurster's blue dihydrochloride, 1,4-benzenediamine, n,n,n',n'-tetramethyl-, dihydrochloride, 1,4-benzenediamine, n1,n1,n4,n4-tetramethyl-, hydrochloride 1:2, 1-n,1-n,4-n,4-n-tetramethylbenzene-1,4-diamine dihydrochloride, wursters reagent
PubChem CID 71561
IUPAC Name 1-N,1-N,4-N,4-N-tetramethylbenzene-1,4-diamine;dihydrochloride
SMILES CN(C)C1=CC=C(C=C1)N(C)C.Cl.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 220°C to 224°C
Color Beige to Gray
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula C6H4[N(CH3)2]2·2HCl
Quantity 100 g
Beilstein 13, 74
Merck Index 10, 9052
Solubility Information Solubility in water: ca 50g/L
Formula Weight 237.17
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • N,N,N`,N`-Tetramethyl-p-phenylenediamine dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.