missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N,N,N',N'-Tetramethylethylenediamine, 99%, for biochemistry, For electrophoresis

£27.14 - £1268.31

Chemical Identifiers

CAS 110-18-9
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.21
MDL Number MFCD00008335
InChI Key KWYHDKDOAIKMQN-UHFFFAOYSA-N
Synonym temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
PubChem CID 8037
ChEBI CHEBI:32850
IUPAC Name N,N,N',N'-tetramethylethane-1,2-diamine
SMILES CN(C)CCN(C)C
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10760102
View Documents
Thermo Scientific Chemicals
420580050
5 g Glass bottle
£27.14
5g
Please to purchase this item. Need a web account? Register with us today!
10168180
View Documents Promotion Details
Thermo Scientific Chemicals
420580500
50 g Glass bottle
£37.59
50g
Please to purchase this item. Need a web account? Register with us today!
10137560
View Documents
Thermo Scientific Chemicals
420581000
100 g Glass bottle
£63.01
100g
Please to purchase this item. Need a web account? Register with us today!
10005660
View Documents
Thermo Scientific Chemicals
420585000
500 g Glass bottle
£297.48
500g
Please to purchase this item. Need a web account? Register with us today!
10398863
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
420580025
2.5 kg Plastic Bottle
£1268.31
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécification

Identifiants chimiques

110-18-9
116.21
KWYHDKDOAIKMQN-UHFFFAOYSA-N
8037
N,N,N',N'-tetramethylethane-1,2-diamine
C6H16N2
MFCD00008335
temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
CHEBI:32850
CN(C)CCN(C)C

Spécification

110-18-9
100.0
Colorless to Yellow
120.0°C to 122.0°C
Authentic
C6H16N2
1.4169 to 1.4189
04, 250
15, 9277
temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
KWYHDKDOAIKMQN-UHFFFAOYSA-N
N,N,N',N'-tetramethylethane-1,2-diamine
8037
116.21
Biochemistry
N,N,N',N'-Tetramethylethylenediamine, 99%
98.5
-55.0°C
0.7700g/mL
17°C
98.5% min. (GC)
(CH3)2NCH2CH2N(CH3)2
MFCD00008335
02,403; 03,284; 04,485; 05,652; 06,576; 07,358; 11,340; 12,477; 14,68; 15,63; 16,55
0.77
(10% in water) Clear and no haze
CN(C)CCN(C)C
116.21
CHEBI:32850
99%
Liquid
Vidéos
Sécurité et traitement

Sécurité et traitement

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 203-744-6

missing translation for 'rtecsNumber' : KV7175000

missing translation for 'tsca' : TSCA

FDS
Documents

Documents

Certificats

RUO – Research Use Only