missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N,N',N'-Tetramethylethylenediamine, 99%, extra pure, Thermo Scientific Chemicals
£17.10 - £214.00
Chemical Identifiers
CAS | 110-18-9 |
---|---|
Molecular Formula | C6H16N2 |
Molecular Weight (g/mol) | 116.21 |
InChI Key | KWYHDKDOAIKMQN-UHFFFAOYSA-N |
Synonym | temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl |
PubChem CID | 8037 |
ChEBI | CHEBI:32850 |
IUPAC Name | N,N,N',N'-tetramethylethane-1,2-diamine |
SMILES | CN(C)CCN(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10265090
|
Thermo Scientific Acros
138450500 |
50 mL | Glass bottle |
£17.10
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10103000
|
Thermo Scientific Acros
138455000 |
500 mL | Glass bottle |
£59.10
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11905191
|
Thermo Scientific Acros
138450025 |
2.5 L | Glass bottle |
£214.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-18-9 | |
116.21 | |
temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl | |
CHEBI:32850 | |
CN(C)CCN(C)C |
C6H16N2 | |
KWYHDKDOAIKMQN-UHFFFAOYSA-N | |
8037 | |
N,N,N',N'-tetramethylethane-1,2-diamine |
Specifications
110-18-9 | |
0.7700g/mL | |
17°C | |
98.5% min. (GC) | |
(CH3)2NCH2CH2N(CH3)2 | |
04,25 | |
15,9277 | |
temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl | |
KWYHDKDOAIKMQN-UHFFFAOYSA-N | |
N,N,N',N'-tetramethylethane-1,2-diamine | |
8037 | |
116.21 | |
Extra Pure | |
N,N,N',N'-Tetramethylethylenediamine |
-55.0°C | |
120.0°C to 122.0°C | |
Authentic | |
C6H16N2 | |
1.4169 to 1.4189 | |
02,403; 03,284; 04,485; 05,652; 06,576; 07,358; 11,340; 12,477; 14,68; 15,63; 16,55 | |
0.77 | |
Solubility in water: miscible. Other solubilities: most organic solvents | |
CN(C)CCN(C)C | |
116.21 | |
CHEBI:32850 | |
99% | |
Liquid |
Safety and Handling
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-744-6
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- N,N,N',N'-Tetramethylethylenediamine
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
RUO – Research Use Only