Learn More
N,N,N',N'-Tetramethylethylenediamine, 99%, Thermo Scientific Chemicals
£21.00 - £218.09
Chemical Identifiers
CAS | 110-18-9 |
---|---|
Molecular Formula | C6H16N2 |
Molecular Weight (g/mol) | 116.208 |
MDL Number | MFCD00008335 |
InChI Key | KWYHDKDOAIKMQN-UHFFFAOYSA-N |
Synonym | temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl |
PubChem CID | 8037 |
ChEBI | CHEBI:32850 |
IUPAC Name | N,N,N',N'-tetramethylethane-1,2-diamine |
SMILES | CN(C)CCN(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11424214
|
Thermo Scientific Alfa Aesar
A12536.AE |
100 mL |
£21.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11434214
|
Thermo Scientific Alfa Aesar
A12536.AP |
500 mL |
£55.38
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11444214
|
Thermo Scientific Alfa Aesar
A12536.0F |
2500 mL |
£218.09
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N,N,N',N'-Tetramethylethylenediamine is used with ammonium persulfate to catalyze the polymerization of acrylamide and bis-acrylamide to get polyacrylamide gels. It is also used for the separation of proteins or nucleic acids. Further, it is employed as a ligand for metal ions like zinc and copper. It is actively involved in the formation of anionic organometallic complex.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N,N′,N′-Tetramethylethylenediamine is used with ammonium persulfate to catalyze the polymerization of acrylamide and bis-acrylamide to get polyacrylamide gels. It is also used for the separation of proteins or nucleic acids. Further, it is employed as a ligand for metal ions like zinc and copper. It is actively involved in the formation of anionic organometallic complex.
Solubility
Miscible with water.
Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, carbon dioxide and copper.
Chemical Identifiers
110-18-9 | |
116.208 | |
KWYHDKDOAIKMQN-UHFFFAOYSA-N | |
8037 | |
N,N,N',N'-tetramethylethane-1,2-diamine |
C6H16N2 | |
MFCD00008335 | |
temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl | |
CHEBI:32850 | |
CN(C)CCN(C)C |
Specifications
110-18-9 | |
0.775 | |
18°C (62°F) | |
C6H16N2 | |
1.418 | |
UN2372 | |
Air sensitive; Hygroscopic | |
Miscible with water. | |
CN(C)CCN(C)C | |
116.208 | |
CHEBI:32850 | |
99% | |
N,N,N',N'-Tetramethylethylenediamine |
°C to 55°C | |
120°C to 122°C | |
99% | |
(CH3)2NCH2CH2N(CH3)2 | |
MFCD00008335 | |
1732991 | |
temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl | |
KWYHDKDOAIKMQN-UHFFFAOYSA-N | |
N,N,N',N'-tetramethylethane-1,2-diamine | |
8037 | |
116.21 | |
Liquid |
Safety and Handling
GHS H Statement
H225-H314-H302-H332
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302+H332-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 1,2-DI-(DIMETHYLAMINO)ETHANE
missing translation for 'einecsNumber' : 203-744-6
missing translation for 'rtecsNumber' : KV7175000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : Hygroscopic
Recommended Storage : Ambient temperatures; Store under Argon
- N,N,N',N'-Tetramethylethylenediamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : 1,2-Bis(dimethylamino)ethane
RUO – Research Use Only