Learn More
N,N,N'-Trimethylethylenediamine, 97%, Thermo Scientific Chemicals
£38.80 - £410.00
Chemical Identifiers
CAS | 142-25-6 |
---|---|
Molecular Formula | C5H14N2 |
Molecular Weight (g/mol) | 102.18 |
MDL Number | MFCD00014874 |
InChI Key | HVOYZOQVDYHUPF-UHFFFAOYSA-N |
Synonym | n,n,n'-trimethylethylenediamine, n1,n1,n2-trimethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n'-trimethyl, 2-dimethylamino ethyl methyl amine, n,n,n-trimethylethylenediamine, ethylenediamine, n,n,n'-trimethyl, n1,n1,n2-trimethyl-1,2-ethanediamine, 1,2-ethanediamine, n1,n1,n2-trimethyl, dimethyl 2-methylamino ethyl amine, n,n,n'-trimethylethane-1,2-diamine |
PubChem CID | 67338 |
IUPAC Name | N,N',N'-trimethylethane-1,2-diamine |
SMILES | CNCCN(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10394542
|
Thermo Scientific
366910050 |
5 g | Glass bottle |
£38.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10406252
|
Thermo Scientific Acros
366910250 |
25 g | Glass bottle |
£135.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10307773
|
Thermo Scientific Acros
366911000 |
100 g | Glass bottle |
£410.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
142-25-6 | |
102.18 | |
HVOYZOQVDYHUPF-UHFFFAOYSA-N | |
67338 | |
CNCCN(C)C |
C5H14N2 | |
MFCD00014874 | |
n,n,n'-trimethylethylenediamine, n1,n1,n2-trimethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n'-trimethyl, 2-dimethylamino ethyl methyl amine, n,n,n-trimethylethylenediamine, ethylenediamine, n,n,n'-trimethyl, n1,n1,n2-trimethyl-1,2-ethanediamine, 1,2-ethanediamine, n1,n1,n2-trimethyl, dimethyl 2-methylamino ethyl amine, n,n,n'-trimethylethane-1,2-diamine | |
N,N',N'-trimethylethane-1,2-diamine |
Specifications
142-25-6 | |
100.0 | |
0.8000g/mL | |
9°C | |
96% min. (GC) | |
(CH3)2NCH2CH2NHCH3 | |
MFCD00014874 | |
0.8 | |
HVOYZOQVDYHUPF-UHFFFAOYSA-N | |
N,N',N'-trimethylethane-1,2-diamine | |
67338 | |
97% | |
N, N, N'-Trimethylethylenediamine |
96.0 | |
Colorless | |
116°C to 118°C | |
Authentic | |
C5H14N2 | |
1.4185 to 1.4205 | |
04, I, 415 | |
n,n,n'-trimethylethylenediamine, n1,n1,n2-trimethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n'-trimethyl, 2-dimethylamino ethyl methyl amine, n,n,n-trimethylethylenediamine, ethylenediamine, n,n,n'-trimethyl, n1,n1,n2-trimethyl-1,2-ethanediamine, 1,2-ethanediamine, n1,n1,n2-trimethyl, dimethyl 2-methylamino ethyl amine, n,n,n'-trimethylethane-1,2-diamine | |
CNCCN(C)C | |
102.18 | |
102.18 | |
Liquid |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Causes serious eye damage.
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 205-529-2
- N,N,N'-Trimethylethylenediamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only