Learn More
N,N,N'-Trimethylethylenediamine, 96%, Thermo Scientific Chemicals
Description
N,N,N'-Trimethylethylenediamine is used as a precursor in the synthesis of a novel gemini surfactant viz. N,N-dimethyl-N-{2-[N'-methyl-N'-(3-sulfopropyl)-alkylammonium]ethyl}-1-alkylammonium bromides. It is also used as an amine component for in-situ formation of alpha-amino alkoxides. Further, it is involved in the synthesis of N,N'-dimethyl-N'-(2-hydroxy-aethyl)-aethylendiamin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Used as precursor in synthesis of novel gemini surfactant viz. N,N-dimethyl-N-{2-[N'-methyl-N'-(3-sulfopropyl)-alkylammonium]ethyl}-1-alkylammonium bromides
- Used as amine component for in-situ formation of alpha-amino alkoxides Involved in synthesis of N,N'-dimethyl-N'-(2-hydroxy-aethyl)-aethylendiamin
Specifications
Specifications
Density | 0.804 g/mL |
Boiling Point | 116°C to 118°C |
Flash Point | 9°C (49°F) |
Odor | Amine-like |
Linear Formula | (CH3)2NCH2CH2NHCH3 |
Refractive Index | 1.419 |
Quantity | 25 g |
UN Number | UN2733 |
Formula Weight | 102.18 |
Percent Purity | 98% |
Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.