missing translation for 'onlineSavingsMsg'
Learn More

N-(n-Octyl)-n-pentylamine, 98+%, Thermo Scientific™

Product Code. 11369525
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6835-13-8
Molecular Formula C17H15ClNO4
Molecular Weight (g/mol) 332.76
MDL Number MFCD03093075
InChI Key HEFAXWHJDHPLEJ-UHFFFAOYNA-M
Synonym n-n-octyl-n-pentylamine, pentyloctylamine, octyl pentyl amine, n-n-octyl-n-pentylamine, 98+%
PubChem CID 20069586
SMILES CC(OC1=CC=C(C=C1)C(=O)NC1=CC=C(Cl)C=C1C)C([O-])=O

Specifications

Refractive Index 1.437
Quantity 1 g
UN Number UN2735
Beilstein 7524567
Sensitivity Air Sensitive
IUPAC Name 2-{4-[(4-chloro-2-methylphenyl)carbamoyl]phenoxy}propanoate
Formula Weight 199.38
Percent Purity ≥98%
Chemical Name or Material N-(n-Octyl)-n-pentylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'