missing translation for 'onlineSavingsMsg'
Learn More
n-Nitrosodimethylamine, SPEX CertiPrep™ Product Code.: 12989314
compliance-icons compliance-icons compliance-icons

n-Nitrosodimethylamine, SPEX CertiPrep™

Product Code. 12989314 Shop All SPEX Certiprep Products
1 mL
Click to view available options
Quantity:
1 mL
This item is not returnable. View return policy

Product Code. 12989314

Brand: SPEX Certiprep S2815

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
N-Nitrosodimethylamine62-75-91000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 62-75-9, 67-56-1
Packaging 2mL Amber Prescored Glass Ampule
CAS Min % 0.1, 99.9
Quantity 1 mL
CAS Max % 0.1, 99.9
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
Molecular Formula C2H6N2O
InChI Key UMFJAHHVKNCGLG-UHFFFAOYSA-N
SMILES CN(C)N=O
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
IUPAC Name N,N-dimethylnitrous amide
Molecular Weight (g/mol) 74.083
PubChem CID 6124
ChEBI CHEBI:35807
Type Certified Reference Material
Solvent or Matrix Methanol-P&T
Chemical Name or Material n-Nitrosodimethylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N-Nitrosodimethylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 1B
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H350-May cause cancer.
  • H370-Causes damage to organs.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : methanol, dimethylnitrosoamine

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-Nitrosodimethylamine, SPEX CertiPrep™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.