missing translation for 'onlineSavingsMsg'
Learn More
n-Nonane, 99%, Thermo Scientific Chemicals Product Code.: 11433247

n-Nonane, 99%, Thermo Scientific Chemicals

Product Code. 11433247
500 mL
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 11433247

Brand: Thermo Scientific Alfa Aesar A16177.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Nonane is used in organic synthesis, as a solvent, as a distillation chaser and as a fuel additive. It is also used in biodegradable detergent. It is an active ingredient of such petroleum fractions as varnish makers and painter naphtha, 140 flash, stoddard solvents and in gasoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nonane is used in organic synthesis, as a solvent, as a distillation chaser and as a fuel additive. It is also used in biodegradable detergent. It is an active ingredient of such petroleum fractions as varnish makers and painter naphtha, 140 flash, stoddard solvents and in gasoline.

Solubility
Miscible with ethanol, ether, acetone, benzene, chloroform and hydrogen peroxide. Immiscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-84-2
Molecular Formula C9H20
Molecular Weight (g/mol) 128.259
MDL Number MFCD00009574
InChI Key BKIMMITUMNQMOS-UHFFFAOYSA-N
Synonym n-nonane, shellsol 140, nonyl hydride, nonan, heptane, ethyl, unii-t9w3vh6g10, ccris 6081, hsdb 107, dsstox_cid_5796, nonane, analytical standard
PubChem CID 8141
ChEBI CHEBI:32892
IUPAC Name nonane
SMILES CCCCCCCCC

Specifications

Melting Point -53°C
Density 0.718
Boiling Point 150°C to 152°C
Flash Point 31°C (87°F)
Linear Formula CH3(CH2)7CH3
Refractive Index 1.4054
Quantity 500 mL
UN Number UN1920
Beilstein 1696917
Solubility Information Miscible with ethanol,ether,acetone,benzene,chloroform and hydrogen peroxide. Immiscible with water.
Formula Weight 128.26
Percent Purity 99%
Chemical Name or Material n-Nonane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10246357 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberA16177.AP
Lot Number10223919 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16177.AP,A16177.AE
Lot NumberG28Q01A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16177.AP,A16177.AE
Lot Number10146767 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16177.AP,A16177.AE
Lot Number10161731 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16177.AP,A16177.AE
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Nonane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contains : Nonane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-Nonane, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.